Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1109152-1109425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
UniGene
Hs.408708
Gene Chromosome Location
Chr.19: 1107634-1174283 on Build GRCh38
Transcripts
NM_001100122.1 NM_014963.2 XM_011527804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2813460 c.3342C>A p.P1114P Chr.19: 1109352-1109352 on Build GRCh38
6538967 c.3415G>C p.E1139Q Chr.19: 1109199-1109199 on Build GRCh38
1611519 c.3300G>A p.R1100R Chr.19: 1109340-1109340 on Build GRCh38
4937772 c.3249G>A p.A1083A Chr.19: 1109391-1109391 on Build GRCh38
5458703 c.3289A>G p.S1097G Chr.19: 1109405-1109405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11669443 Chr.19: 1109216-1109216 on Build GRCh38
rs374055757 Chr.19: 1109282-1109282 on Build GRCh38
rs745595357 Chr.19: 1109304-1109304 on Build GRCh38
rs369926315 Chr.19: 1109281-1109281 on Build GRCh38
rs1017192552 Chr.19: 1109229-1109229 on Build GRCh38
rs768765538 Chr.19: 1109243-1109243 on Build GRCh38
rs749100891 Chr.19: 1109336-1109336 on Build GRCh38
rs1005746646 Chr.19: 1109226-1109226 on Build GRCh38
rs1003948930 Chr.19: 1109247-1109247 on Build GRCh38
rs769994144 Chr.19: 1109404-1109404 on Build GRCh38
rs376859092 Chr.19: 1109228-1109228 on Build GRCh38
rs1038736914 Chr.19: 1109181-1109181 on Build GRCh38
rs113371241 Chr.19: 1109235-1109235 on Build GRCh38
rs750855118 Chr.19: 1109207-1109207 on Build GRCh38
rs781001227 Chr.19: 1109391-1109391 on Build GRCh38
rs770087414 Chr.19: 1109316-1109316 on Build GRCh38
rs750763443 Chr.19: 1109269-1109269 on Build GRCh38
rs944363655 Chr.19: 1109173-1109173 on Build GRCh38
rs745695455 Chr.19: 1109223-1109223 on Build GRCh38
rs748958999 Chr.19: 1109215-1109215 on Build GRCh38
rs1054165729 Chr.19: 1109361-1109361 on Build GRCh38
rs773945716 Chr.19: 1109301-1109302 on Build GRCh38
rs988605209 Chr.19: 1109279-1109279 on Build GRCh38
rs778550295 Chr.19: 1109301-1109301 on Build GRCh38
rs376424828 Chr.19: 1109294-1109294 on Build GRCh38
rs776620612 Chr.19: 1109249-1109249 on Build GRCh38
rs564235160 Chr.19: 1109346-1109346 on Build GRCh38
rs765583526 Chr.19: 1109264-1109264 on Build GRCh38
rs746807840 Chr.19: 1109374-1109374 on Build GRCh38
rs764154895 Chr.19: 1109203-1109203 on Build GRCh38
rs775500991 Chr.19: 1109230-1109230 on Build GRCh38
rs373078345 Chr.19: 1109313-1109313 on Build GRCh38
rs750062195 Chr.19: 1109363-1109363 on Build GRCh38
rs780733055 Chr.19: 1109209-1109209 on Build GRCh38
rs763428414 Chr.19: 1109278-1109278 on Build GRCh38
rs768512524 Chr.19: 1109308-1109308 on Build GRCh38
rs2159133 Chr.19: 1109214-1109214 on Build GRCh38
rs758034628 Chr.19: 1109364-1109364 on Build GRCh38
rs201689400 Chr.19: 1109293-1109293 on Build GRCh38
rs370823591 Chr.19: 1109305-1109305 on Build GRCh38
rs778648591 Chr.19: 1109217-1109217 on Build GRCh38
rs891557765 Chr.19: 1109382-1109382 on Build GRCh38
rs746941279 Chr.19: 1109307-1109307 on Build GRCh38
rs1054220924 Chr.19: 1109212-1109212 on Build GRCh38
rs900116746 Chr.19: 1109232-1109232 on Build GRCh38
rs11880694 Chr.19: 1109323-1109323 on Build GRCh38
rs751149593 Chr.19: 1109343-1109343 on Build GRCh38
rs752094834 Chr.19: 1109210-1109210 on Build GRCh38
rs531590571 Chr.19: 1109376-1109376 on Build GRCh38
rs201860402 Chr.19: 1109206-1109206 on Build GRCh38
rs766755426 Chr.19: 1109331-1109331 on Build GRCh38
rs756764477 Chr.19: 1109352-1109352 on Build GRCh38
rs766732664 Chr.19: 1109280-1109280 on Build GRCh38
rs775382883 Chr.19: 1109197-1109197 on Build GRCh38
rs753296059 Chr.19: 1109349-1109349 on Build GRCh38
rs544310103 Chr.19: 1109341-1109341 on Build GRCh38
rs760820981 Chr.19: 1109198-1109198 on Build GRCh38
rs779768771 Chr.19: 1109306-1109306 on Build GRCh38
rs1029929911 Chr.19: 1109350-1109350 on Build GRCh38
rs755657269 Chr.19: 1109211-1109211 on Build GRCh38
rs372665804 Chr.19: 1109221-1109221 on Build GRCh38
rs773486167 Chr.19: 1109406-1109406 on Build GRCh38
rs1024192896 Chr.19: 1109234-1109234 on Build GRCh38
rs760595780 Chr.19: 1109238-1109238 on Build GRCh38
rs973429843 Chr.19: 1109286-1109286 on Build GRCh38
rs779866050 Chr.19: 1109365-1109365 on Build GRCh38
rs772037840 Chr.19: 1109171-1109171 on Build GRCh38
rs181011896 Chr.19: 1109227-1109227 on Build GRCh38
rs762092380 Chr.19: 1109254-1109254 on Build GRCh38
rs748177445 Chr.19: 1109392-1109392 on Build GRCh38
rs758930002 Chr.19: 1109208-1109208 on Build GRCh38
rs754084380 Chr.19: 1109204-1109204 on Build GRCh38
rs759447854 Chr.19: 1109170-1109170 on Build GRCh38
rs945853025 Chr.19: 1109403-1109403 on Build GRCh38
rs747688500 Chr.19: 1109365-1109365 on Build GRCh38

View Full Product Details