Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29940581-29940842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRN1
Entrez Gene ID
9805
Gene Name
secernin 1
Gene Aliases
SES1
UniGene
Hs.520740
Gene Chromosome Location
Chr.7: 29920103-29990289 on Build GRCh38
Transcripts
NM_001145513.1 NM_001145514.1 NM_001145515.1 NM_014766.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4933261 c.668C>G p.S223C Chr.7: 29940753-29940753 on Build GRCh38
5062420 c.715G>A p.E239K Chr.7: 29940766-29940766 on Build GRCh38
1329876 c.751G>C p.D251H Chr.7: 29940730-29940730 on Build GRCh38
6177686 c.625C>G p.Q209E Chr.7: 29940796-29940796 on Build GRCh38
6177688 c.682C>G p.P228A Chr.7: 29940739-29940739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371750124 Chr.7: 29940714-29940714 on Build GRCh38
rs766957235 Chr.7: 29940769-29940769 on Build GRCh38
rs749264557 Chr.7: 29940704-29940704 on Build GRCh38
rs761270639 Chr.7: 29940767-29940767 on Build GRCh38
rs563879011 Chr.7: 29940799-29940799 on Build GRCh38
rs778949653 Chr.7: 29940712-29940712 on Build GRCh38
rs762692499 Chr.7: 29940752-29940752 on Build GRCh38
rs138423685 Chr.7: 29940708-29940708 on Build GRCh38
rs769256493 Chr.7: 29940800-29940800 on Build GRCh38
rs757611798 Chr.7: 29940672-29940672 on Build GRCh38
rs770795123 Chr.7: 29940729-29940729 on Build GRCh38
rs1004137281 Chr.7: 29940798-29940798 on Build GRCh38
rs747005113 Chr.7: 29940724-29940724 on Build GRCh38
rs764729045 Chr.7: 29940669-29940669 on Build GRCh38
rs373701823 Chr.7: 29940662-29940662 on Build GRCh38
rs375023545 Chr.7: 29940770-29940770 on Build GRCh38
rs917445955 Chr.7: 29940725-29940725 on Build GRCh38
rs367919883 Chr.7: 29940701-29940701 on Build GRCh38
rs927268281 Chr.7: 29940673-29940673 on Build GRCh38
rs762711079 Chr.7: 29940809-29940809 on Build GRCh38
rs777871607 Chr.7: 29940777-29940777 on Build GRCh38
rs773747511 Chr.7: 29940815-29940815 on Build GRCh38
rs758928262 Chr.7: 29940746-29940746 on Build GRCh38
rs756679527 Chr.7: 29940693-29940693 on Build GRCh38
rs772015371 Chr.7: 29940650-29940650 on Build GRCh38
rs768704750 Chr.7: 29940634-29940634 on Build GRCh38
rs765937757 Chr.7: 29940661-29940661 on Build GRCh38
rs775045104 Chr.7: 29940751-29940751 on Build GRCh38
rs768359884 Chr.7: 29940813-29940813 on Build GRCh38
rs767965419 Chr.7: 29940679-29940679 on Build GRCh38
rs775052891 Chr.7: 29940808-29940808 on Build GRCh38
rs1051583231 Chr.7: 29940697-29940697 on Build GRCh38
rs777556223 Chr.7: 29940780-29940780 on Build GRCh38
rs774205258 Chr.7: 29940642-29940642 on Build GRCh38
rs969639720 Chr.7: 29940796-29940796 on Build GRCh38
rs375510462 Chr.7: 29940613-29940613 on Build GRCh38
rs375395964 Chr.7: 29940816-29940816 on Build GRCh38
rs563763444 Chr.7: 29940785-29940785 on Build GRCh38
rs769325526 Chr.7: 29940747-29940747 on Build GRCh38
rs779015479 Chr.7: 29940776-29940776 on Build GRCh38
rs201681112 Chr.7: 29940637-29940637 on Build GRCh38
rs143901036 Chr.7: 29940716-29940716 on Build GRCh38
rs770886787 Chr.7: 29940786-29940786 on Build GRCh38
rs751039693 Chr.7: 29940685-29940685 on Build GRCh38
rs750761419 Chr.7: 29940766-29940766 on Build GRCh38
rs768053514 Chr.7: 29940765-29940765 on Build GRCh38
rs780941872 Chr.7: 29940792-29940792 on Build GRCh38
rs150361714 Chr.7: 29940771-29940771 on Build GRCh38
rs900154751 Chr.7: 29940803-29940803 on Build GRCh38
rs865981587 Chr.7: 29940610-29940610 on Build GRCh38
rs771892542 Chr.7: 29940715-29940715 on Build GRCh38
rs747861164 Chr.7: 29940646-29940646 on Build GRCh38
rs773178357 Chr.7: 29940660-29940660 on Build GRCh38
rs776209901 Chr.7: 29940737-29940737 on Build GRCh38

View Full Product Details