Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35752035-35752260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN37
Entrez Gene ID
55957
Gene Name
lin-37 DREAM MuvB core complex component
Gene Aliases
F25965, ZK418.4, lin-37
UniGene
Hs.529100
Gene Chromosome Location
Chr.19: 35748361-35754519 on Build GRCh38
Transcripts
NM_019104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7222580 c.106G>A p.D36N Chr.19: 35752247-35752247 on Build GRCh38
7230667 c.45G>A p.M15I Chr.19: 35752186-35752186 on Build GRCh38
1285929 c.46G>A p.V16I Chr.19: 35752187-35752187 on Build GRCh38
4590601 c.100C>T p.H34Y Chr.19: 35752241-35752241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530617326 Chr.19: 35752232-35752232 on Build GRCh38
rs747842588 Chr.19: 35752221-35752221 on Build GRCh38
rs960708344 Chr.19: 35752151-35752151 on Build GRCh38
rs1014787965 Chr.19: 35752110-35752110 on Build GRCh38
rs11878987 Chr.19: 35752097-35752097 on Build GRCh38
rs201196330 Chr.19: 35752210-35752210 on Build GRCh38
rs746490365 Chr.19: 35752236-35752236 on Build GRCh38
rs767651009 Chr.19: 35752178-35752178 on Build GRCh38
rs774625507 Chr.19: 35752165-35752165 on Build GRCh38
rs368488713 Chr.19: 35752142-35752142 on Build GRCh38
rs1004310566 Chr.19: 35752072-35752072 on Build GRCh38
rs548169729 Chr.19: 35752111-35752111 on Build GRCh38
rs905023179 Chr.19: 35752054-35752054 on Build GRCh38
rs1027877971 Chr.19: 35752186-35752186 on Build GRCh38
rs776450687 Chr.19: 35752134-35752134 on Build GRCh38
rs1002334305 Chr.19: 35752122-35752122 on Build GRCh38
rs565863907 Chr.19: 35752126-35752126 on Build GRCh38
rs771002888 Chr.19: 35752137-35752160 on Build GRCh38
rs778497912 Chr.19: 35752213-35752213 on Build GRCh38
rs937817952 Chr.19: 35752086-35752086 on Build GRCh38
rs887378917 Chr.19: 35752062-35752062 on Build GRCh38
rs761872739 Chr.19: 35752172-35752172 on Build GRCh38
rs1014924343 Chr.19: 35752113-35752113 on Build GRCh38
rs200797120 Chr.19: 35752222-35752222 on Build GRCh38
rs796989672 Chr.19: 35752187-35752188 on Build GRCh38
rs562059547 Chr.19: 35752093-35752093 on Build GRCh38
rs770903533 Chr.19: 35752237-35752237 on Build GRCh38
rs170758 Chr.19: 35752188-35752188 on Build GRCh38
rs754303935 Chr.19: 35752204-35752204 on Build GRCh38
rs895866599 Chr.19: 35752098-35752098 on Build GRCh38
rs764277636 Chr.19: 35752157-35752157 on Build GRCh38
rs766965615 Chr.19: 35752196-35752196 on Build GRCh38
rs756723431 Chr.19: 35752194-35752194 on Build GRCh38
rs762834489 Chr.19: 35752138-35752138 on Build GRCh38
rs913798209 Chr.19: 35752197-35752197 on Build GRCh38

View Full Product Details