Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10557438-10557681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRI1
Entrez Gene ID
65095
Gene Name
KRI1 homolog
Gene Aliases
-
UniGene
Hs.709967
Gene Chromosome Location
Chr.19: 10553085-10566031 on Build GRCh38
Transcripts
NM_023008.3 XM_011528190.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
710177 c.1576C>G p.P526A Chr.19: 10557611-10557611 on Build GRCh38
7269685 c.1555G>A p.E519K Chr.19: 10557632-10557632 on Build GRCh38
4696740 c.1515G>A p.T505T Chr.19: 10557672-10557672 on Build GRCh38
1162542 c.1538A>G p.Y513C Chr.19: 10557649-10557649 on Build GRCh38
143572 c.1505_1506GG>AA p.G502E Chr.19: 10557681-10557682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144674130 Chr.19: 10557564-10557564 on Build GRCh38
rs764777753 Chr.19: 10557609-10557609 on Build GRCh38
rs768297032 Chr.19: 10557584-10557585 on Build GRCh38
rs986932804 Chr.19: 10557519-10557519 on Build GRCh38
rs914338568 Chr.19: 10557615-10557615 on Build GRCh38
rs781756291 Chr.19: 10557509-10557509 on Build GRCh38
rs755547586 Chr.19: 10557509-10557537 on Build GRCh38
rs920844662 Chr.19: 10557579-10557579 on Build GRCh38
rs368692136 Chr.19: 10557633-10557633 on Build GRCh38
rs971403663 Chr.19: 10557478-10557478 on Build GRCh38
rs756796833 Chr.19: 10557502-10557502 on Build GRCh38
rs150629769 Chr.19: 10557632-10557632 on Build GRCh38
rs942327753 Chr.19: 10557561-10557561 on Build GRCh38
rs778573526 Chr.19: 10557614-10557614 on Build GRCh38
rs757405353 Chr.19: 10557643-10557643 on Build GRCh38
rs370952722 Chr.19: 10557589-10557589 on Build GRCh38
rs376337650 Chr.19: 10557626-10557626 on Build GRCh38
rs748731788 Chr.19: 10557514-10557514 on Build GRCh38
rs200104114 Chr.19: 10557503-10557503 on Build GRCh38
rs745638378 Chr.19: 10557553-10557553 on Build GRCh38
rs764100625 Chr.19: 10557592-10557592 on Build GRCh38
rs200865594 Chr.19: 10557568-10557568 on Build GRCh38
rs749276934 Chr.19: 10557637-10557637 on Build GRCh38
rs932991819 Chr.19: 10557621-10557623 on Build GRCh38
rs1023363095 Chr.19: 10557462-10557463 on Build GRCh38
rs755390588 Chr.19: 10557618-10557618 on Build GRCh38
rs934484845 Chr.19: 10557528-10557528 on Build GRCh38
rs774654041 Chr.19: 10557550-10557550 on Build GRCh38
rs779575386 Chr.19: 10557582-10557583 on Build GRCh38
rs901536989 Chr.19: 10557620-10557620 on Build GRCh38
rs142840947 Chr.19: 10557617-10557617 on Build GRCh38
rs775967399 Chr.19: 10557605-10557605 on Build GRCh38
rs534747076 Chr.19: 10557474-10557474 on Build GRCh38
rs771018153 Chr.19: 10557544-10557544 on Build GRCh38
rs754941063 Chr.19: 10557602-10557602 on Build GRCh38
rs930849419 Chr.19: 10557593-10557593 on Build GRCh38
rs536447005 Chr.19: 10557540-10557540 on Build GRCh38
rs546700841 Chr.19: 10557494-10557494 on Build GRCh38
rs753323702 Chr.19: 10557644-10557644 on Build GRCh38
rs756205930 Chr.19: 10557527-10557527 on Build GRCh38
rs1047874784 Chr.19: 10557598-10557598 on Build GRCh38
rs775398165 Chr.19: 10557588-10557588 on Build GRCh38
rs558559887 Chr.19: 10557621-10557621 on Build GRCh38
rs748915084 Chr.19: 10557583-10557584 on Build GRCh38
rs201666146 Chr.19: 10557541-10557541 on Build GRCh38
rs747948683 Chr.19: 10557642-10557643 on Build GRCh38
rs750587508 Chr.19: 10557597-10557597 on Build GRCh38
rs755445537 Chr.19: 10557507-10557507 on Build GRCh38
rs570531161 Chr.19: 10557472-10557472 on Build GRCh38

View Full Product Details