Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135665254-135665491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCN9
Entrez Gene ID
392399
Gene Name
lipocalin 9
Gene Aliases
9230102I19Rik, HEL129
UniGene
Hs.532679
Gene Chromosome Location
Chr.9: 135663322-135670731 on Build GRCh38
Transcripts
NM_001001676.1 XM_017014712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4774607 c.429+9C>A p.? Chr.9: 135665375-135665375 on Build GRCh38
1213181 c.427G>A p.A143T Chr.9: 135665364-135665364 on Build GRCh38
1106667 c.335C>T p.S112L Chr.9: 135665272-135665272 on Build GRCh38
7449386 c.402C>A p.H134Q Chr.9: 135665339-135665339 on Build GRCh38
162127 c.418+8C>T p.? Chr.9: 135665363-135665363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766745432 Chr.9: 135665386-135665386 on Build GRCh38
rs561054922 Chr.9: 135665322-135665322 on Build GRCh38
rs954289313 Chr.9: 135665377-135665377 on Build GRCh38
rs780633998 Chr.9: 135665329-135665329 on Build GRCh38
rs557129387 Chr.9: 135665273-135665273 on Build GRCh38
rs533723011 Chr.9: 135665341-135665341 on Build GRCh38
rs762374126 Chr.9: 135665315-135665315 on Build GRCh38
rs193005693 Chr.9: 135665347-135665347 on Build GRCh38
rs773765170 Chr.9: 135665381-135665381 on Build GRCh38
rs1028331939 Chr.9: 135665369-135665369 on Build GRCh38
rs531042875 Chr.9: 135665429-135665429 on Build GRCh38
rs762427566 Chr.9: 135665367-135665367 on Build GRCh38
rs925589011 Chr.9: 135665397-135665397 on Build GRCh38
rs1044229269 Chr.9: 135665439-135665439 on Build GRCh38
rs766577451 Chr.9: 135665327-135665327 on Build GRCh38
rs775067076 Chr.9: 135665280-135665280 on Build GRCh38
rs940093401 Chr.9: 135665415-135665415 on Build GRCh38
rs367648675 Chr.9: 135665342-135665342 on Build GRCh38
rs492708 Chr.9: 135665410-135665410 on Build GRCh38
rs768316985 Chr.9: 135665363-135665363 on Build GRCh38
rs542930081 Chr.9: 135665321-135665321 on Build GRCh38
rs61742118 Chr.9: 135665348-135665348 on Build GRCh38
rs755100258 Chr.9: 135665438-135665438 on Build GRCh38
rs752155644 Chr.9: 135665291-135665291 on Build GRCh38
rs370267826 Chr.9: 135665325-135665325 on Build GRCh38
rs199765604 Chr.9: 135665289-135665289 on Build GRCh38
rs916993477 Chr.9: 135665434-135665434 on Build GRCh38
rs927185174 Chr.9: 135665471-135665471 on Build GRCh38
rs750695452 Chr.9: 135665324-135665324 on Build GRCh38
rs532246424 Chr.9: 135665364-135665364 on Build GRCh38
rs575312352 Chr.9: 135665279-135665279 on Build GRCh38
rs754097969 Chr.9: 135665404-135665404 on Build GRCh38
rs755097116 Chr.9: 135665330-135665330 on Build GRCh38
rs748921454 Chr.9: 135665353-135665353 on Build GRCh38
rs761004290 Chr.9: 135665382-135665382 on Build GRCh38
rs151041662 Chr.9: 135665379-135665379 on Build GRCh38
rs746978499 Chr.9: 135665337-135665337 on Build GRCh38
rs374188235 Chr.9: 135665331-135665331 on Build GRCh38
rs988569931 Chr.9: 135665425-135665425 on Build GRCh38

View Full Product Details