Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77908818-77909043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHTF2
Entrez Gene ID
57157
Gene Name
putative homeodomain transcription factor 2
Gene Aliases
-
UniGene
Hs.203965
Gene Chromosome Location
Chr.7: 77798792-77957504 on Build GRCh38
Transcripts
NM_001127357.1 NM_001127358.1 NM_001127359.1 NM_001127360.1 NM_020432.4 XM_005250508.4 XM_005250509.4 XM_006716063.3 XM_011516422.2 XM_011516423.2 XM_011516424.2 XM_017012458.1 XM_017012459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220492 c.605G>T p.R202I Chr.7: 77908952-77908952 on Build GRCh38
6745681 c.611+6_611+7insT p.? Chr.7: 77908964-77908965 on Build GRCh38
6745676 c.395T>C p.L132P Chr.7: 77908844-77908844 on Build GRCh38
1220490 c.512G>A p.G171E Chr.7: 77908859-77908859 on Build GRCh38
485586 c.437G>A p.G146E Chr.7: 77908886-77908886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766452002 Chr.7: 77908896-77908896 on Build GRCh38
rs375153750 Chr.7: 77908839-77908839 on Build GRCh38
rs563308823 Chr.7: 77908985-77908985 on Build GRCh38
rs1050061728 Chr.7: 77909012-77909012 on Build GRCh38
rs967210265 Chr.7: 77908958-77908958 on Build GRCh38
rs765182826 Chr.7: 77908938-77908938 on Build GRCh38
rs566205277 Chr.7: 77908863-77908863 on Build GRCh38
rs746439940 Chr.7: 77908862-77908862 on Build GRCh38
rs369840951 Chr.7: 77908881-77908881 on Build GRCh38
rs36004157 Chr.7: 77908933-77908933 on Build GRCh38
rs750163685 Chr.7: 77908940-77908940 on Build GRCh38
rs958606802 Chr.7: 77908935-77908935 on Build GRCh38
rs776342579 Chr.7: 77908873-77908873 on Build GRCh38
rs200528794 Chr.7: 77908931-77908931 on Build GRCh38
rs904639352 Chr.7: 77908942-77908942 on Build GRCh38
rs770461087 Chr.7: 77908882-77908882 on Build GRCh38
rs773668092 Chr.7: 77908890-77908890 on Build GRCh38
rs977668899 Chr.7: 77908976-77908976 on Build GRCh38
rs758219535 Chr.7: 77908943-77908943 on Build GRCh38
rs1055069995 Chr.7: 77908855-77908855 on Build GRCh38
rs923023898 Chr.7: 77908978-77908978 on Build GRCh38
rs754640699 Chr.7: 77909008-77909008 on Build GRCh38
rs764129651 Chr.7: 77908934-77908934 on Build GRCh38
rs763396047 Chr.7: 77908894-77908894 on Build GRCh38
rs779772271 Chr.7: 77908843-77908843 on Build GRCh38
rs774635847 Chr.7: 77908915-77908915 on Build GRCh38
rs757146800 Chr.7: 77908937-77908937 on Build GRCh38
rs746663725 Chr.7: 77908997-77908997 on Build GRCh38
rs779783072 Chr.7: 77908946-77908946 on Build GRCh38

View Full Product Details