Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24417967-24418192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRS2
Entrez Gene ID
57380
Gene Name
MRS2, magnesium transporter
Gene Aliases
HPT, MRS2L
UniGene
Hs.658612
Gene Chromosome Location
Chr.6: 24402908-24426194 on Build GRCh38
Transcripts
NM_001286264.1 NM_001286265.1 NM_001286266.1 NM_020662.3 NR_104423.1 XM_005249242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1215530 c.931C>T p.R311C Chr.6: 24418178-24418178 on Build GRCh38
3860730 c.883C>T p.L295L Chr.6: 24418130-24418130 on Build GRCh38
2856608 c.901C>T p.R301* Chr.6: 24418148-24418148 on Build GRCh38
3948837 c.880G>A p.E294K Chr.6: 24418118-24418118 on Build GRCh38
5830357 c.882G>A p.E294E Chr.6: 24418120-24418120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745499349 Chr.6: 24418043-24418043 on Build GRCh38
rs772917568 Chr.6: 24418060-24418060 on Build GRCh38
rs1030325081 Chr.6: 24418005-24418005 on Build GRCh38
rs776645690 Chr.6: 24418077-24418077 on Build GRCh38
rs989828098 Chr.6: 24418074-24418074 on Build GRCh38
rs372577192 Chr.6: 24418065-24418065 on Build GRCh38
rs886214086 Chr.6: 24418149-24418149 on Build GRCh38
rs2793422 Chr.6: 24418120-24418120 on Build GRCh38
rs774415035 Chr.6: 24418072-24418072 on Build GRCh38
rs537841265 Chr.6: 24418080-24418080 on Build GRCh38
rs201847935 Chr.6: 24418171-24418171 on Build GRCh38
rs768519267 Chr.6: 24418058-24418061 on Build GRCh38
rs747825363 Chr.6: 24418049-24418049 on Build GRCh38
rs375281394 Chr.6: 24418045-24418045 on Build GRCh38
rs781343643 Chr.6: 24418136-24418136 on Build GRCh38
rs142761910 Chr.6: 24418147-24418147 on Build GRCh38
rs757652949 Chr.6: 24418126-24418126 on Build GRCh38
rs776253911 Chr.6: 24418173-24418173 on Build GRCh38
rs765335854 Chr.6: 24418098-24418098 on Build GRCh38
rs752725510 Chr.6: 24418105-24418105 on Build GRCh38
rs781720865 Chr.6: 24418038-24418038 on Build GRCh38
rs764348521 Chr.6: 24418116-24418116 on Build GRCh38
rs746587042 Chr.6: 24418170-24418170 on Build GRCh38
rs760511113 Chr.6: 24418073-24418073 on Build GRCh38
rs981110178 Chr.6: 24418159-24418159 on Build GRCh38
rs554666112 Chr.6: 24418111-24418111 on Build GRCh38
rs768284407 Chr.6: 24418039-24418039 on Build GRCh38
rs772904013 Chr.6: 24418161-24418161 on Build GRCh38
rs761911440 Chr.6: 24418122-24418123 on Build GRCh38
rs891279601 Chr.6: 24418014-24418014 on Build GRCh38
rs190278916 Chr.6: 24418055-24418055 on Build GRCh38
rs766282348 Chr.6: 24418076-24418076 on Build GRCh38
rs747627568 Chr.6: 24418160-24418160 on Build GRCh38
rs778611295 Chr.6: 24418148-24418148 on Build GRCh38
rs746407611 Chr.6: 24418146-24418146 on Build GRCh38

View Full Product Details