Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12647192-12647427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6706663 c.2882G>A p.R961H Chr.19: 12647271-12647271 on Build GRCh38
3422438 c.2826C>T p.F942F Chr.19: 12647327-12647327 on Build GRCh38
4846193 c.2884G>A p.E962K Chr.19: 12647269-12647269 on Build GRCh38
40833 c.2897G>A p.R966K Chr.19: 12647256-12647256 on Build GRCh38
991359 c.2898G>A p.R966R Chr.19: 12647255-12647255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544567546 Chr.19: 12647279-12647279 on Build GRCh38
rs765084966 Chr.19: 12647348-12647348 on Build GRCh38
rs370276057 Chr.19: 12647272-12647272 on Build GRCh38
rs373821522 Chr.19: 12647217-12647217 on Build GRCh38
rs750173812 Chr.19: 12647271-12647271 on Build GRCh38
rs753776387 Chr.19: 12647339-12647339 on Build GRCh38
rs771210668 Chr.19: 12647292-12647292 on Build GRCh38
rs774791244 Chr.19: 12647235-12647236 on Build GRCh38
rs771288611 Chr.19: 12647224-12647224 on Build GRCh38
rs559212808 Chr.19: 12647369-12647369 on Build GRCh38
rs764063183 Chr.19: 12647263-12647263 on Build GRCh38
rs1004058684 Chr.19: 12647390-12647390 on Build GRCh38
rs377438700 Chr.19: 12647264-12647264 on Build GRCh38
rs139041112 Chr.19: 12647307-12647307 on Build GRCh38
rs148108322 Chr.19: 12647291-12647291 on Build GRCh38
rs957664522 Chr.19: 12647391-12647391 on Build GRCh38
rs536242066 Chr.19: 12647251-12647251 on Build GRCh38
rs977787836 Chr.19: 12647222-12647222 on Build GRCh38
rs374333674 Chr.19: 12647375-12647375 on Build GRCh38
rs369122358 Chr.19: 12647371-12647371 on Build GRCh38
rs1057516897 Chr.19: 12647234-12647234 on Build GRCh38
rs779879898 Chr.19: 12647368-12647368 on Build GRCh38
rs768233248 Chr.19: 12647289-12647289 on Build GRCh38
rs761870446 Chr.19: 12647340-12647340 on Build GRCh38
rs752840068 Chr.19: 12647245-12647245 on Build GRCh38
rs746571094 Chr.19: 12647286-12647286 on Build GRCh38
rs370579499 Chr.19: 12647351-12647351 on Build GRCh38
rs749331220 Chr.19: 12647393-12647393 on Build GRCh38
rs757072948 Chr.19: 12647269-12647269 on Build GRCh38
rs990275905 Chr.19: 12647395-12647396 on Build GRCh38
rs775663639 Chr.19: 12647325-12647325 on Build GRCh38
rs750273286 Chr.19: 12647350-12647350 on Build GRCh38
rs774443107 Chr.19: 12647226-12647226 on Build GRCh38
rs1024929407 Chr.19: 12647363-12647363 on Build GRCh38
rs148080695 Chr.19: 12647239-12647239 on Build GRCh38
rs759814559 Chr.19: 12647308-12647308 on Build GRCh38
rs142797984 Chr.19: 12647270-12647270 on Build GRCh38
rs780794113 Chr.19: 12647372-12647372 on Build GRCh38
rs906602699 Chr.19: 12647370-12647370 on Build GRCh38
rs200431869 Chr.19: 12647253-12647253 on Build GRCh38
rs771531945 Chr.19: 12647380-12647380 on Build GRCh38
rs759744566 Chr.19: 12647238-12647238 on Build GRCh38
rs1047812725 Chr.19: 12647352-12647352 on Build GRCh38
rs760762187 Chr.19: 12647333-12647333 on Build GRCh38
rs377567861 Chr.19: 12647310-12647310 on Build GRCh38
rs764116542 Chr.19: 12647338-12647338 on Build GRCh38

View Full Product Details