Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49133096-49133325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIA3
Entrez Gene ID
8541
Gene Name
PTPRF interacting protein alpha 3
Gene Aliases
LPNA3
UniGene
Hs.413748
Gene Chromosome Location
Chr.19: 49119389-49151030 on Build GRCh38
Transcripts
NM_003660.3 NR_103842.1 XM_017027407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282566 c.1023G>A p.R341R Chr.19: 49133144-49133144 on Build GRCh38
7158400 c.1038G>A p.K346K Chr.19: 49133248-49133248 on Build GRCh38
6390111 c.986T>A p.L329Q Chr.19: 49133107-49133107 on Build GRCh38
7078969 c.1013C>T p.S338L Chr.19: 49133134-49133134 on Build GRCh38
135843 c.1026+2T>G p.? Chr.19: 49133149-49133149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970582331 Chr.19: 49133198-49133198 on Build GRCh38
rs928601813 Chr.19: 49133178-49133178 on Build GRCh38
rs769860160 Chr.19: 49133181-49133181 on Build GRCh38
rs775077562 Chr.19: 49133171-49133171 on Build GRCh38
rs755686191 Chr.19: 49133155-49133155 on Build GRCh38
rs200420754 Chr.19: 49133222-49133222 on Build GRCh38
rs375459144 Chr.19: 49133251-49133251 on Build GRCh38
rs751653959 Chr.19: 49133190-49133190 on Build GRCh38
rs780062608 Chr.19: 49133263-49133263 on Build GRCh38
rs777508004 Chr.19: 49133156-49133156 on Build GRCh38
rs949483145 Chr.19: 49133217-49133217 on Build GRCh38
rs773470922 Chr.19: 49133185-49133185 on Build GRCh38
rs772055292 Chr.19: 49133165-49133165 on Build GRCh38
rs980596523 Chr.19: 49133208-49133208 on Build GRCh38
rs746756493 Chr.19: 49133299-49133299 on Build GRCh38
rs368168621 Chr.19: 49133129-49133129 on Build GRCh38
rs781353888 Chr.19: 49133149-49133149 on Build GRCh38
rs753608660 Chr.19: 49133220-49133220 on Build GRCh38
rs369298116 Chr.19: 49133150-49133150 on Build GRCh38
rs748786925 Chr.19: 49133157-49133157 on Build GRCh38
rs759969142 Chr.19: 49133192-49133192 on Build GRCh38
rs763626555 Chr.19: 49133218-49133218 on Build GRCh38
rs371870415 Chr.19: 49133207-49133207 on Build GRCh38
rs766505149 Chr.19: 49133189-49133189 on Build GRCh38
rs1019581491 Chr.19: 49133303-49133303 on Build GRCh38
rs778504908 Chr.19: 49133228-49133228 on Build GRCh38
rs760548169 Chr.19: 49133174-49133174 on Build GRCh38
rs368474557 Chr.19: 49133186-49133186 on Build GRCh38
rs778987116 Chr.19: 49133164-49133164 on Build GRCh38
rs889263094 Chr.19: 49133270-49133270 on Build GRCh38
rs751641430 Chr.19: 49133155-49133161 on Build GRCh38
rs141749268 Chr.19: 49133135-49133135 on Build GRCh38
rs552464930 Chr.19: 49133307-49133307 on Build GRCh38
rs762425392 Chr.19: 49133230-49133230 on Build GRCh38
rs753111277 Chr.19: 49133153-49133153 on Build GRCh38
rs745372754 Chr.19: 49133242-49133242 on Build GRCh38
rs926361039 Chr.19: 49133213-49133213 on Build GRCh38
rs756553394 Chr.19: 49133215-49133215 on Build GRCh38
rs770608454 Chr.19: 49133160-49133160 on Build GRCh38

View Full Product Details