Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1491295-1491522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REEP6
Entrez Gene ID
92840
Gene Name
receptor accessory protein 6
Gene Aliases
C19orf32, DP1L1, TB2L1, Yip2f
UniGene
Hs.76277
Gene Chromosome Location
Chr.19: 1491166-1497925 on Build GRCh38
Transcripts
NM_138393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991340 c.245A>G p.K82R Chr.19: 1491312-1491312 on Build GRCh38
4791614 c.52G>T p.V18F Chr.19: 1491321-1491321 on Build GRCh38
1189756 c.89G>C p.G30A Chr.19: 1491358-1491358 on Build GRCh38
6991339 c.43A>G p.R15G Chr.19: 1491312-1491312 on Build GRCh38
7280729 c.56C>T p.T19I Chr.19: 1491325-1491325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372605180 Chr.19: 1491394-1491394 on Build GRCh38
rs989753923 Chr.19: 1491415-1491415 on Build GRCh38
rs901177993 Chr.19: 1491364-1491364 on Build GRCh38
rs561512625 Chr.19: 1491379-1491379 on Build GRCh38
rs779919665 Chr.19: 1491328-1491328 on Build GRCh38
rs566007230 Chr.19: 1491398-1491398 on Build GRCh38
rs992442625 Chr.19: 1491502-1491502 on Build GRCh38
rs758779051 Chr.19: 1491479-1491479 on Build GRCh38
rs759940069 Chr.19: 1491435-1491435 on Build GRCh38
rs1042504192 Chr.19: 1491495-1491495 on Build GRCh38
rs772454499 Chr.19: 1491403-1491413 on Build GRCh38
rs959713014 Chr.19: 1491497-1491497 on Build GRCh38
rs781223650 Chr.19: 1491357-1491357 on Build GRCh38
rs1010095005 Chr.19: 1491405-1491405 on Build GRCh38
rs750986429 Chr.19: 1491413-1491413 on Build GRCh38
rs945565832 Chr.19: 1491443-1491443 on Build GRCh38
rs965484445 Chr.19: 1491365-1491365 on Build GRCh38
rs927004828 Chr.19: 1491488-1491488 on Build GRCh38
rs769995368 Chr.19: 1491378-1491378 on Build GRCh38
rs930190815 Chr.19: 1491335-1491335 on Build GRCh38
rs915452887 Chr.19: 1491426-1491426 on Build GRCh38
rs906696876 Chr.19: 1491504-1491504 on Build GRCh38
rs139331241 Chr.19: 1491346-1491346 on Build GRCh38
rs369183038 Chr.19: 1491360-1491360 on Build GRCh38
rs549769578 Chr.19: 1491314-1491314 on Build GRCh38
rs539208242 Chr.19: 1491496-1491496 on Build GRCh38
rs956136622 Chr.19: 1491395-1491395 on Build GRCh38
rs746879220 Chr.19: 1491331-1491331 on Build GRCh38
rs528912161 Chr.19: 1491390-1491390 on Build GRCh38
rs979828126 Chr.19: 1491463-1491463 on Build GRCh38
rs1021872268 Chr.19: 1491432-1491432 on Build GRCh38

View Full Product Details