Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72821916-72822141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIBF1
Entrez Gene ID
10464
Gene Name
progesterone immunomodulatory binding factor 1
Gene Aliases
C13orf24, CEP90, PIBF
UniGene
Hs.441926
Gene Chromosome Location
Chr.13: 72782059-73016454 on Build GRCh38
Transcripts
NM_006346.2 XM_011534881.2 XM_011534882.2 XM_011534884.2 XM_011534885.2 XM_011534886.2 XM_017020350.1 XM_017020351.1 XM_017020352.1 XM_017020353.1 XM_017020354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5984353 c.784G>C p.E262Q Chr.13: 72821960-72821960 on Build GRCh38
7163313 c.793G>A p.D265N Chr.13: 72821969-72821969 on Build GRCh38
5478580 c.771T>C p.G257G Chr.13: 72821947-72821947 on Build GRCh38
6198748 c.806+9T>C p.? Chr.13: 72821991-72821991 on Build GRCh38
6390408 c.766C>A p.Q256K Chr.13: 72821942-72821942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542089852 Chr.13: 72821981-72821981 on Build GRCh38
rs760173850 Chr.13: 72821969-72821969 on Build GRCh38
rs767948792 Chr.13: 72822034-72822034 on Build GRCh38
rs199982144 Chr.13: 72821964-72821964 on Build GRCh38
rs770366184 Chr.13: 72821972-72821972 on Build GRCh38
rs766604368 Chr.13: 72821955-72821955 on Build GRCh38
rs764417242 Chr.13: 72821996-72821996 on Build GRCh38
rs748308148 Chr.13: 72822026-72822039 on Build GRCh38
rs776303882 Chr.13: 72821975-72821975 on Build GRCh38
rs930398058 Chr.13: 72821947-72821947 on Build GRCh38
rs889919215 Chr.13: 72822105-72822105 on Build GRCh38
rs756155608 Chr.13: 72821945-72821945 on Build GRCh38
rs901450948 Chr.13: 72822041-72822041 on Build GRCh38
rs549677199 Chr.13: 72822108-72822108 on Build GRCh38
rs762305892 Chr.13: 72822028-72822028 on Build GRCh38
rs759148284 Chr.13: 72821982-72821982 on Build GRCh38
rs748083623 Chr.13: 72821963-72821963 on Build GRCh38
rs114942614 Chr.13: 72822085-72822085 on Build GRCh38
rs751981029 Chr.13: 72822013-72822013 on Build GRCh38
rs957093403 Chr.13: 72822075-72822075 on Build GRCh38
rs1011445609 Chr.13: 72822106-72822106 on Build GRCh38
rs780265946 Chr.13: 72821954-72821954 on Build GRCh38
rs376173304 Chr.13: 72821965-72821965 on Build GRCh38
rs778893360 Chr.13: 72821962-72821962 on Build GRCh38
rs754605020 Chr.13: 72822015-72822015 on Build GRCh38

View Full Product Details