Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18118905-18119134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
UniGene
Hs.462390
Gene Chromosome Location
Chr.17: 18108706-18180227 on Build GRCh38
Transcripts
NM_016239.3 XM_011523918.2 XM_017024714.1 XM_017024715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364124 c.271C>T p.R91C Chr.17: 18119071-18119071 on Build GRCh38
6556212 c.185G>T p.G62V Chr.17: 18118985-18118985 on Build GRCh38
4528224 c.151G>A p.G51S Chr.17: 18118951-18118951 on Build GRCh38
704838 c.166G>A p.A56T Chr.17: 18118966-18118966 on Build GRCh38
1679617 c.138G>T p.K46N Chr.17: 18118938-18118938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747477704 Chr.17: 18118956-18118956 on Build GRCh38
rs745436438 Chr.17: 18119049-18119049 on Build GRCh38
rs753710398 Chr.17: 18119093-18119093 on Build GRCh38
rs569331416 Chr.17: 18118952-18118952 on Build GRCh38
rs760706490 Chr.17: 18119086-18119086 on Build GRCh38
rs770052089 Chr.17: 18118970-18118970 on Build GRCh38
rs748573149 Chr.17: 18118966-18118966 on Build GRCh38
rs765000005 Chr.17: 18119111-18119111 on Build GRCh38
rs781715805 Chr.17: 18119051-18119051 on Build GRCh38
rs752534089 Chr.17: 18119029-18119029 on Build GRCh38
rs748660803 Chr.17: 18119054-18119054 on Build GRCh38
rs776139570 Chr.17: 18118948-18118949 on Build GRCh38
rs766321344 Chr.17: 18118995-18118995 on Build GRCh38
rs774557155 Chr.17: 18119071-18119071 on Build GRCh38
rs755813626 Chr.17: 18118938-18118938 on Build GRCh38
rs751394944 Chr.17: 18118922-18118922 on Build GRCh38
rs761353840 Chr.17: 18119100-18119100 on Build GRCh38
rs778258793 Chr.17: 18119043-18119043 on Build GRCh38
rs752587062 Chr.17: 18118929-18118929 on Build GRCh38
rs888945293 Chr.17: 18119030-18119030 on Build GRCh38
rs775508030 Chr.17: 18119076-18119076 on Build GRCh38
rs753738434 Chr.17: 18119022-18119022 on Build GRCh38
rs763250174 Chr.17: 18118978-18118978 on Build GRCh38
rs759755751 Chr.17: 18119072-18119072 on Build GRCh38
rs773648511 Chr.17: 18119063-18119063 on Build GRCh38
rs754996079 Chr.17: 18118924-18118924 on Build GRCh38
rs767619427 Chr.17: 18119016-18119016 on Build GRCh38
rs760881582 Chr.17: 18119075-18119076 on Build GRCh38
rs549511151 Chr.17: 18118941-18118941 on Build GRCh38
rs748973041 Chr.17: 18118951-18118951 on Build GRCh38
rs571886925 Chr.17: 18119064-18119064 on Build GRCh38
rs759523877 Chr.17: 18119015-18119015 on Build GRCh38
rs374370560 Chr.17: 18119074-18119074 on Build GRCh38
rs933253559 Chr.17: 18118975-18118975 on Build GRCh38
rs923156097 Chr.17: 18118928-18118928 on Build GRCh38
rs371411411 Chr.17: 18119007-18119007 on Build GRCh38
rs373294263 Chr.17: 18119069-18119069 on Build GRCh38
rs188609046 Chr.17: 18119050-18119050 on Build GRCh38
rs942397992 Chr.17: 18119053-18119053 on Build GRCh38
rs199689781 Chr.17: 18119089-18119089 on Build GRCh38
rs369442352 Chr.17: 18118965-18118965 on Build GRCh38
rs767388920 Chr.17: 18118927-18118927 on Build GRCh38
rs200827453 Chr.17: 18119092-18119092 on Build GRCh38
rs755902122 Chr.17: 18119034-18119034 on Build GRCh38
rs778504453 Chr.17: 18118953-18118953 on Build GRCh38
rs753378643 Chr.17: 18119040-18119040 on Build GRCh38
rs201749134 Chr.17: 18119035-18119035 on Build GRCh38

View Full Product Details