Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158721284-158721535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPID
Entrez Gene ID
5481
Gene Name
peptidylprolyl isomerase D
Gene Aliases
CYP-40, CYPD
UniGene
Hs.183958
Gene Chromosome Location
Chr.4: 158709127-158723400 on Build GRCh38
Transcripts
NM_005038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3760459 c.145C>T p.R49C Chr.4: 158721424-158721424 on Build GRCh38
7171182 c.170G>C p.G57A Chr.4: 158721399-158721399 on Build GRCh38
6753458 c.177A>G p.G59G Chr.4: 158721392-158721392 on Build GRCh38
7192027 c.213C>T p.C71C Chr.4: 158721356-158721356 on Build GRCh38
4475559 c.199C>T p.H67Y Chr.4: 158721370-158721370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201322183 Chr.4: 158721435-158721435 on Build GRCh38
rs771239741 Chr.4: 158721417-158721417 on Build GRCh38
rs186494786 Chr.4: 158721396-158721396 on Build GRCh38
rs746957379 Chr.4: 158721477-158721477 on Build GRCh38
rs2070631 Chr.4: 158721424-158721424 on Build GRCh38
rs752223056 Chr.4: 158721465-158721465 on Build GRCh38
rs764081687 Chr.4: 158721335-158721335 on Build GRCh38
rs773751438 Chr.4: 158721410-158721410 on Build GRCh38
rs145953224 Chr.4: 158721455-158721455 on Build GRCh38
rs528791439 Chr.4: 158721500-158721500 on Build GRCh38
rs766369113 Chr.4: 158721486-158721490 on Build GRCh38
rs553055223 Chr.4: 158721373-158721373 on Build GRCh38
rs555339594 Chr.4: 158721339-158721339 on Build GRCh38
rs559747481 Chr.4: 158721485-158721485 on Build GRCh38
rs761783799 Chr.4: 158721434-158721434 on Build GRCh38
rs369501739 Chr.4: 158721423-158721423 on Build GRCh38
rs199670143 Chr.4: 158721346-158721346 on Build GRCh38
rs746024673 Chr.4: 158721504-158721504 on Build GRCh38
rs762916156 Chr.4: 158721475-158721475 on Build GRCh38
rs774537416 Chr.4: 158721322-158721322 on Build GRCh38
rs746598178 Chr.4: 158721356-158721356 on Build GRCh38
rs761231161 Chr.4: 158721387-158721387 on Build GRCh38
rs200023691 Chr.4: 158721350-158721350 on Build GRCh38
rs749725944 Chr.4: 158721484-158721484 on Build GRCh38
rs778127676 Chr.4: 158721478-158721478 on Build GRCh38
rs142096418 Chr.4: 158721449-158721449 on Build GRCh38
rs749592443 Chr.4: 158721411-158721411 on Build GRCh38
rs759518763 Chr.4: 158721333-158721333 on Build GRCh38
rs750293636 Chr.4: 158721438-158721438 on Build GRCh38
rs764980502 Chr.4: 158721345-158721345 on Build GRCh38
rs147585130 Chr.4: 158721431-158721431 on Build GRCh38
rs753704667 Chr.4: 158721336-158721336 on Build GRCh38
rs763194324 Chr.4: 158721314-158721314 on Build GRCh38
rs774428808 Chr.4: 158721421-158721421 on Build GRCh38
rs766477218 Chr.4: 158721318-158721318 on Build GRCh38
rs756732759 Chr.4: 158721466-158721466 on Build GRCh38
rs762653393 Chr.4: 158721448-158721448 on Build GRCh38
rs529213590 Chr.4: 158721437-158721437 on Build GRCh38
rs148824218 Chr.4: 158721370-158721370 on Build GRCh38
rs572328740 Chr.4: 158721348-158721348 on Build GRCh38
rs754557537 Chr.4: 158721453-158721453 on Build GRCh38
rs751116720 Chr.4: 158721450-158721450 on Build GRCh38

View Full Product Details