Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75399479-75399742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS3
Entrez Gene ID
6188
Gene Name
ribosomal protein S3
Gene Aliases
S3
UniGene
Hs.546286
Gene Chromosome Location
Chr.11: 75399491-75422302 on Build GRCh38
Transcripts
NM_001005.4 NM_001256802.1 NM_001260506.1 NM_001260507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260559 c.3G>T p.M1I Chr.11: 75399550-75399550 on Build GRCh38
5769840 c.7G>T p.V3L Chr.11: 75399554-75399554 on Build GRCh38
4917911 c.11A>G p.Q4R Chr.11: 75399558-75399558 on Build GRCh38
6323157 c.1A>T p.M1L Chr.11: 75399548-75399548 on Build GRCh38
5970380 c.6A>G p.A2A Chr.11: 75399553-75399553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889185769 Chr.11: 75399673-75399673 on Build GRCh38
rs7131286 Chr.11: 75399636-75399636 on Build GRCh38
rs763765147 Chr.11: 75399522-75399522 on Build GRCh38
rs772964979 Chr.11: 75399558-75399558 on Build GRCh38
rs372675348 Chr.11: 75399513-75399513 on Build GRCh38
rs368313317 Chr.11: 75399623-75399623 on Build GRCh38
rs751211175 Chr.11: 75399505-75399505 on Build GRCh38
rs759481359 Chr.11: 75399514-75399514 on Build GRCh38
rs768505909 Chr.11: 75399620-75399620 on Build GRCh38
rs775565062 Chr.11: 75399544-75399544 on Build GRCh38
rs994818734 Chr.11: 75399663-75399663 on Build GRCh38
rs967151226 Chr.11: 75399518-75399518 on Build GRCh38
rs747257091 Chr.11: 75399512-75399512 on Build GRCh38
rs17881757 Chr.11: 75399531-75399531 on Build GRCh38
rs759858656 Chr.11: 75399559-75399559 on Build GRCh38
rs17880657 Chr.11: 75399532-75399532 on Build GRCh38
rs1028181771 Chr.11: 75399672-75399672 on Build GRCh38
rs1018961707 Chr.11: 75399618-75399618 on Build GRCh38
rs1033354284 Chr.11: 75399562-75399562 on Build GRCh38
rs751616244 Chr.11: 75399593-75399593 on Build GRCh38
rs920676502 Chr.11: 75399668-75399668 on Build GRCh38
rs771091990 Chr.11: 75399541-75399541 on Build GRCh38
rs531798394 Chr.11: 75399537-75399537 on Build GRCh38
rs866356929 Chr.11: 75399684-75399684 on Build GRCh38
rs773735949 Chr.11: 75399546-75399546 on Build GRCh38
rs987773457 Chr.11: 75399610-75399610 on Build GRCh38
rs755426109 Chr.11: 75399528-75399528 on Build GRCh38
rs772184770 Chr.11: 75399510-75399510 on Build GRCh38
rs553767972 Chr.11: 75399519-75399519 on Build GRCh38
rs374130065 Chr.11: 75399596-75399596 on Build GRCh38
rs780007016 Chr.11: 75399611-75399611 on Build GRCh38
rs375300446 Chr.11: 75399507-75399507 on Build GRCh38
rs766698183 Chr.11: 75399524-75399524 on Build GRCh38
rs3133187 Chr.11: 75399545-75399545 on Build GRCh38
rs111486717 Chr.11: 75399580-75399580 on Build GRCh38
rs1001912436 Chr.11: 75399554-75399554 on Build GRCh38
rs755961163 Chr.11: 75399604-75399604 on Build GRCh38
rs983625523 Chr.11: 75399682-75399682 on Build GRCh38
rs35773854 Chr.11: 75399650-75399651 on Build GRCh38
rs540509110 Chr.11: 75399511-75399511 on Build GRCh38
rs570846657 Chr.11: 75399619-75399619 on Build GRCh38
rs17879266 Chr.11: 75399583-75399583 on Build GRCh38
rs528006215 Chr.11: 75399584-75399584 on Build GRCh38
rs201273793 Chr.11: 75399530-75399530 on Build GRCh38
rs1046077337 Chr.11: 75399552-75399552 on Build GRCh38
rs748246278 Chr.11: 75399509-75399509 on Build GRCh38
rs930793085 Chr.11: 75399536-75399536 on Build GRCh38
rs745637925 Chr.11: 75399542-75399542 on Build GRCh38
rs781172604 Chr.11: 75399673-75399673 on Build GRCh38
rs369334575 Chr.11: 75399553-75399553 on Build GRCh38
rs202233761 Chr.11: 75399500-75399500 on Build GRCh38
rs746159771 Chr.11: 75399601-75399601 on Build GRCh38
rs112375628 Chr.11: 75399578-75399578 on Build GRCh38
rs767095371 Chr.11: 75399556-75399556 on Build GRCh38
rs200915680 Chr.11: 75399523-75399523 on Build GRCh38
rs765762609 Chr.11: 75399543-75399543 on Build GRCh38
rs17884475 Chr.11: 75399700-75399700 on Build GRCh38
rs373443936 Chr.11: 75399565-75399565 on Build GRCh38
rs939710830 Chr.11: 75399551-75399551 on Build GRCh38
rs777625112 Chr.11: 75399577-75399578 on Build GRCh38
rs35515336 Chr.11: 75399504-75399504 on Build GRCh38
rs756496152 Chr.11: 75399506-75399506 on Build GRCh38
rs183314111 Chr.11: 75399598-75399598 on Build GRCh38
rs755479260 Chr.11: 75399508-75399508 on Build GRCh38
rs201527751 Chr.11: 75399625-75399625 on Build GRCh38
rs764195666 Chr.11: 75399585-75399585 on Build GRCh38

View Full Product Details