Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47836262-47836507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC46
Entrez Gene ID
90506
Gene Name
leucine rich repeat containing 46
Gene Aliases
-
UniGene
Hs.130767
Gene Chromosome Location
Chr.17: 47831627-47837713 on Build GRCh38
Transcripts
NM_033413.3 XM_005257775.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2697009 c.593G>A p.R198Q Chr.17: 47836473-47836473 on Build GRCh38
6428364 c.475C>G p.P159A Chr.17: 47836355-47836355 on Build GRCh38
4699114 c.534G>A p.E178E Chr.17: 47836414-47836414 on Build GRCh38
980538 c.574G>A p.G192S Chr.17: 47836454-47836454 on Build GRCh38
2697008 c.592C>T p.R198* Chr.17: 47836472-47836472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139048679 Chr.17: 47836395-47836395 on Build GRCh38
rs755779904 Chr.17: 47836459-47836459 on Build GRCh38
rs11654648 Chr.17: 47836287-47836287 on Build GRCh38
rs763977340 Chr.17: 47836429-47836429 on Build GRCh38
rs764100433 Chr.17: 47836406-47836429 on Build GRCh38
rs757050286 Chr.17: 47836437-47836437 on Build GRCh38
rs374240152 Chr.17: 47836451-47836451 on Build GRCh38
rs201474905 Chr.17: 47836405-47836405 on Build GRCh38
rs761726518 Chr.17: 47836433-47836435 on Build GRCh38
rs751287685 Chr.17: 47836334-47836334 on Build GRCh38
rs760911937 Chr.17: 47836386-47836386 on Build GRCh38
rs771263564 Chr.17: 47836482-47836482 on Build GRCh38
rs756965289 Chr.17: 47836345-47836345 on Build GRCh38
rs765326581 Chr.17: 47836310-47836310 on Build GRCh38
rs760473322 Chr.17: 47836292-47836292 on Build GRCh38
rs780657901 Chr.17: 47836353-47836353 on Build GRCh38
rs758314001 Chr.17: 47836316-47836316 on Build GRCh38
rs778380141 Chr.17: 47836478-47836478 on Build GRCh38
rs370207682 Chr.17: 47836430-47836430 on Build GRCh38
rs752713225 Chr.17: 47836422-47836422 on Build GRCh38
rs745510729 Chr.17: 47836359-47836359 on Build GRCh38
rs144936742 Chr.17: 47836473-47836473 on Build GRCh38
rs377582072 Chr.17: 47836363-47836363 on Build GRCh38
rs776755227 Chr.17: 47836394-47836394 on Build GRCh38
rs750110260 Chr.17: 47836449-47836449 on Build GRCh38
rs147341805 Chr.17: 47836393-47836393 on Build GRCh38
rs781010052 Chr.17: 47836438-47836438 on Build GRCh38
rs772650862 Chr.17: 47836376-47836376 on Build GRCh38
rs373525707 Chr.17: 47836288-47836288 on Build GRCh38
rs149897709 Chr.17: 47836404-47836404 on Build GRCh38
rs777036776 Chr.17: 47836484-47836484 on Build GRCh38
rs376286427 Chr.17: 47836464-47836464 on Build GRCh38
rs765176245 Chr.17: 47836457-47836458 on Build GRCh38
rs773719057 Chr.17: 47836382-47836382 on Build GRCh38
rs200098111 Chr.17: 47836375-47836375 on Build GRCh38
rs574769677 Chr.17: 47836428-47836428 on Build GRCh38
rs779708459 Chr.17: 47836373-47836373 on Build GRCh38
rs902711920 Chr.17: 47836354-47836354 on Build GRCh38
rs777554143 Chr.17: 47836333-47836333 on Build GRCh38
rs748743875 Chr.17: 47836472-47836472 on Build GRCh38
rs16948662 Chr.17: 47836481-47836481 on Build GRCh38
rs952303862 Chr.17: 47836291-47836291 on Build GRCh38
rs776529537 Chr.17: 47836406-47836408 on Build GRCh38

View Full Product Details