Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77340565-77340838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK1
Entrez Gene ID
5058
Gene Name
p21 (RAC1) activated kinase 1
Gene Aliases
PAKalpha
UniGene
Hs.435714
Gene Chromosome Location
Chr.11: 77322015-77474063 on Build GRCh38
Transcripts
NM_001128620.1 NM_002576.4 XM_011545080.2 XM_011545081.2 XM_011545082.2 XM_011545083.2 XM_011545084.2 XM_017017846.1 XM_017017847.1 XM_017017848.1 XM_017017849.1 XM_017017850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6986184 c.1038C>A p.Y346* Chr.11: 77340724-77340724 on Build GRCh38
1659278 c.1071A>C p.T357T Chr.11: 77340691-77340691 on Build GRCh38
5904969 c.1116+8C>G p.? Chr.11: 77340638-77340638 on Build GRCh38
5904968 c.1116+8C>G p.? Chr.11: 77340638-77340638 on Build GRCh38
6914766 c.1006G>A p.V336M Chr.11: 77340756-77340756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112968282 Chr.11: 77340799-77340799 on Build GRCh38
rs762128146 Chr.11: 77340699-77340699 on Build GRCh38
rs774724859 Chr.11: 77340697-77340697 on Build GRCh38
rs766431792 Chr.11: 77340779-77340779 on Build GRCh38
rs755247088 Chr.11: 77340607-77340607 on Build GRCh38
rs2725832 Chr.11: 77340638-77340638 on Build GRCh38
rs51500 Chr.11: 77340775-77340775 on Build GRCh38
rs964510835 Chr.11: 77340791-77340791 on Build GRCh38
rs780335338 Chr.11: 77340600-77340600 on Build GRCh38
rs761228478 Chr.11: 77340777-77340777 on Build GRCh38
rs114957769 Chr.11: 77340757-77340757 on Build GRCh38
rs751827882 Chr.11: 77340639-77340639 on Build GRCh38
rs755300367 Chr.11: 77340801-77340801 on Build GRCh38
rs759227026 Chr.11: 77340641-77340641 on Build GRCh38
rs756082505 Chr.11: 77340599-77340599 on Build GRCh38
rs767864262 Chr.11: 77340596-77340596 on Build GRCh38
rs755351376 Chr.11: 77340671-77340671 on Build GRCh38
rs770407187 Chr.11: 77340633-77340633 on Build GRCh38
rs145870074 Chr.11: 77340650-77340650 on Build GRCh38
rs373412134 Chr.11: 77340601-77340601 on Build GRCh38
rs368535897 Chr.11: 77340814-77340814 on Build GRCh38
rs778887226 Chr.11: 77340619-77340619 on Build GRCh38
rs1031591829 Chr.11: 77340711-77340711 on Build GRCh38
rs753933274 Chr.11: 77340787-77340787 on Build GRCh38
rs558024154 Chr.11: 77340632-77340632 on Build GRCh38
rs534472752 Chr.11: 77340756-77340756 on Build GRCh38
rs1013442369 Chr.11: 77340589-77340589 on Build GRCh38
rs201475016 Chr.11: 77340631-77340631 on Build GRCh38

View Full Product Details