Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127749098-127749371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D3C
Entrez Gene ID
10044
Gene Name
SH2 domain containing 3C
Gene Aliases
CHAT, NSP3, PRO34088, SHEP1
UniGene
Hs.306412
Gene Chromosome Location
Chr.9: 127738317-127778769 on Build GRCh38
Transcripts
NM_001142531.1 NM_001142532.1 NM_001142533.1 NM_001142534.1 NM_001252334.1 NM_005489.3 NM_170600.2 XM_005251639.1 XM_011518114.1 XM_011518115.1 XM_011518117.2 XM_017014174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4370596 c.569G>T p.R190L Chr.9: 127749232-127749232 on Build GRCh38
6777022 c.614T>C p.M205T Chr.9: 127749265-127749265 on Build GRCh38
4370601 c.541G>T p.D181Y Chr.9: 127749260-127749260 on Build GRCh38
6777041 c.865C>T p.R289W Chr.9: 127749281-127749281 on Build GRCh38
6115163 c.609C>T p.V203V Chr.9: 127749270-127749270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377345507 Chr.9: 127749201-127749201 on Build GRCh38
rs756841641 Chr.9: 127749342-127749343 on Build GRCh38
rs528314607 Chr.9: 127749300-127749300 on Build GRCh38
rs999276214 Chr.9: 127749170-127749170 on Build GRCh38
rs754420080 Chr.9: 127749251-127749251 on Build GRCh38
rs201301256 Chr.9: 127749243-127749243 on Build GRCh38
rs922310655 Chr.9: 127749246-127749246 on Build GRCh38
rs767967113 Chr.9: 127749232-127749232 on Build GRCh38
rs761308154 Chr.9: 127749318-127749318 on Build GRCh38
rs765398246 Chr.9: 127749219-127749219 on Build GRCh38
rs546135106 Chr.9: 127749220-127749220 on Build GRCh38
rs373936140 Chr.9: 127749277-127749277 on Build GRCh38
rs143526802 Chr.9: 127749325-127749325 on Build GRCh38
rs146755904 Chr.9: 127749329-127749329 on Build GRCh38
rs748740825 Chr.9: 127749161-127749161 on Build GRCh38
rs768813359 Chr.9: 127749125-127749125 on Build GRCh38
rs774581895 Chr.9: 127749291-127749291 on Build GRCh38
rs150752473 Chr.9: 127749267-127749267 on Build GRCh38
rs758000416 Chr.9: 127749266-127749266 on Build GRCh38
rs200412602 Chr.9: 127749341-127749341 on Build GRCh38
rs62587177 Chr.9: 127749175-127749175 on Build GRCh38
rs1030782156 Chr.9: 127749345-127749345 on Build GRCh38
rs764439822 Chr.9: 127749225-127749225 on Build GRCh38
rs776678290 Chr.9: 127749278-127749278 on Build GRCh38
rs776731089 Chr.9: 127749202-127749202 on Build GRCh38
rs373951203 Chr.9: 127749178-127749178 on Build GRCh38
rs745868179 Chr.9: 127749280-127749280 on Build GRCh38
rs771746208 Chr.9: 127749176-127749176 on Build GRCh38
rs868715534 Chr.9: 127749262-127749262 on Build GRCh38
rs774540514 Chr.9: 127749145-127749145 on Build GRCh38
rs199532954 Chr.9: 127749317-127749317 on Build GRCh38
rs369758471 Chr.9: 127749169-127749169 on Build GRCh38
rs149897538 Chr.9: 127749281-127749281 on Build GRCh38
rs140967477 Chr.9: 127749303-127749303 on Build GRCh38
rs61754496 Chr.9: 127749272-127749272 on Build GRCh38
rs761848242 Chr.9: 127749167-127749167 on Build GRCh38
rs368780382 Chr.9: 127749224-127749224 on Build GRCh38
rs142059103 Chr.9: 127749260-127749260 on Build GRCh38
rs751858191 Chr.9: 127749227-127749227 on Build GRCh38
rs759494296 Chr.9: 127749216-127749216 on Build GRCh38
rs757635271 Chr.9: 127749228-127749228 on Build GRCh38
rs775707255 Chr.9: 127749286-127749286 on Build GRCh38
rs757096056 Chr.9: 127749343-127749343 on Build GRCh38
rs746802045 Chr.9: 127749181-127749181 on Build GRCh38
rs929711989 Chr.9: 127749248-127749248 on Build GRCh38
rs909542294 Chr.9: 127749135-127749135 on Build GRCh38
rs146487344 Chr.9: 127749287-127749287 on Build GRCh38
rs2233503 Chr.9: 127749273-127749273 on Build GRCh38
rs781191281 Chr.9: 127749346-127749346 on Build GRCh38
rs149146405 Chr.9: 127749332-127749332 on Build GRCh38
rs1025092375 Chr.9: 127749159-127749159 on Build GRCh38
rs764245494 Chr.9: 127749289-127749289 on Build GRCh38

View Full Product Details