Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43129624-43129883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTK7
Entrez Gene ID
5754
Gene Name
protein tyrosine kinase 7 (inactive)
Gene Aliases
CCK-4, CCK4
UniGene
Hs.90572
Gene Chromosome Location
Chr.6: 43076268-43161720 on Build GRCh38
Transcripts
NM_001270398.1 NM_002821.4 NM_152880.3 NM_152881.3 NM_152882.3 NR_072997.1 NR_072998.1 XM_011514765.2 XM_011514766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1472002 c.429G>C p.Q143H Chr.6: 43129788-43129788 on Build GRCh38
4718868 c.418A>C p.I140L Chr.6: 43129777-43129777 on Build GRCh38
385191 c.423G>T p.Q141H Chr.6: 43129782-43129782 on Build GRCh38
7234037 c.459T>C p.D153D Chr.6: 43129818-43129818 on Build GRCh38
284360 c.445C>T p.R149C Chr.6: 43129804-43129804 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770967533 Chr.6: 43129786-43129786 on Build GRCh38
rs45624340 Chr.6: 43129705-43129705 on Build GRCh38
rs181689659 Chr.6: 43129805-43129805 on Build GRCh38
rs373711860 Chr.6: 43129681-43129681 on Build GRCh38
rs373247020 Chr.6: 43129791-43129791 on Build GRCh38
rs755297368 Chr.6: 43129843-43129843 on Build GRCh38
rs771968063 Chr.6: 43129679-43129679 on Build GRCh38
rs1007842509 Chr.6: 43129695-43129695 on Build GRCh38
rs565963125 Chr.6: 43129743-43129743 on Build GRCh38
rs374956441 Chr.6: 43129828-43129828 on Build GRCh38
rs981700570 Chr.6: 43129676-43129676 on Build GRCh38
rs762588436 Chr.6: 43129708-43129708 on Build GRCh38
rs776489420 Chr.6: 43129691-43129691 on Build GRCh38
rs1027615214 Chr.6: 43129673-43129673 on Build GRCh38
rs745721306 Chr.6: 43129804-43129804 on Build GRCh38
rs967411821 Chr.6: 43129642-43129642 on Build GRCh38
rs756652293 Chr.6: 43129716-43129716 on Build GRCh38
rs1004526943 Chr.6: 43129732-43129732 on Build GRCh38
rs750842394 Chr.6: 43129711-43129711 on Build GRCh38
rs901726790 Chr.6: 43129813-43129813 on Build GRCh38
rs746854546 Chr.6: 43129779-43129779 on Build GRCh38
rs765038851 Chr.6: 43129701-43129701 on Build GRCh38
rs368364305 Chr.6: 43129767-43129767 on Build GRCh38
rs763634529 Chr.6: 43129812-43129812 on Build GRCh38
rs761075920 Chr.6: 43129827-43129827 on Build GRCh38
rs770572944 Chr.6: 43129688-43129688 on Build GRCh38
rs760684735 Chr.6: 43129685-43129685 on Build GRCh38
rs376958481 Chr.6: 43129790-43129790 on Build GRCh38
rs867468488 Chr.6: 43129766-43129766 on Build GRCh38
rs754288914 Chr.6: 43129746-43129746 on Build GRCh38
rs777844136 Chr.6: 43129769-43129769 on Build GRCh38
rs769096948 Chr.6: 43129662-43129662 on Build GRCh38
rs534888100 Chr.6: 43129859-43129859 on Build GRCh38
rs755172717 Chr.6: 43129750-43129750 on Build GRCh38
rs759105208 Chr.6: 43129696-43129696 on Build GRCh38
rs201897432 Chr.6: 43129795-43129795 on Build GRCh38
rs766652585 Chr.6: 43129737-43129737 on Build GRCh38
rs748278641 Chr.6: 43129678-43129678 on Build GRCh38
rs372365197 Chr.6: 43129757-43129757 on Build GRCh38
rs555656649 Chr.6: 43129710-43129710 on Build GRCh38
rs779147838 Chr.6: 43129752-43129752 on Build GRCh38
rs773981614 Chr.6: 43129814-43129814 on Build GRCh38

View Full Product Details