Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93588161-93588392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2F1
Entrez Gene ID
7025
Gene Name
nuclear receptor subfamily 2 group F member 1
Gene Aliases
BBOAS, BBSOAS, COUP-TFI, EAR-3, EAR3, ERBAL3, NR2F2, SVP44, TCFCOUP1, TFCOUP1
UniGene
Hs.519445
Gene Chromosome Location
Chr.5: 93583337-93594613 on Build GRCh38
Transcripts
NM_005654.5 XM_017009797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5535600 c.772G>A p.E258K Chr.5: 93588225-93588225 on Build GRCh38
235057 c.780C>T p.F260F Chr.5: 93588233-93588233 on Build GRCh38
262462 c.904T>G p.F302V Chr.5: 93588357-93588357 on Build GRCh38
6729171 c.759C>T p.R253R Chr.5: 93588212-93588212 on Build GRCh38
3856903 c.738C>T p.T246T Chr.5: 93588191-93588191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774531602 Chr.5: 93588344-93588344 on Build GRCh38
rs751163796 Chr.5: 93588312-93588312 on Build GRCh38
rs747731110 Chr.5: 93588230-93588230 on Build GRCh38
rs369796105 Chr.5: 93588363-93588363 on Build GRCh38
rs749201458 Chr.5: 93588305-93588305 on Build GRCh38
rs1008298998 Chr.5: 93588282-93588282 on Build GRCh38
rs777533706 Chr.5: 93588264-93588264 on Build GRCh38
rs916298801 Chr.5: 93588191-93588191 on Build GRCh38
rs759492782 Chr.5: 93588311-93588311 on Build GRCh38
rs1019282462 Chr.5: 93588287-93588287 on Build GRCh38
rs780649231 Chr.5: 93588192-93588192 on Build GRCh38
rs770721436 Chr.5: 93588304-93588304 on Build GRCh38
rs750683610 Chr.5: 93588356-93588356 on Build GRCh38
rs140323285 Chr.5: 93588234-93588234 on Build GRCh38
rs768598143 Chr.5: 93588320-93588320 on Build GRCh38
rs891380228 Chr.5: 93588371-93588371 on Build GRCh38
rs778504524 Chr.5: 93588218-93588218 on Build GRCh38
rs587777276 Chr.5: 93588208-93588208 on Build GRCh38
rs995390661 Chr.5: 93588222-93588222 on Build GRCh38
rs746752068 Chr.5: 93588296-93588296 on Build GRCh38
rs767837139 Chr.5: 93588354-93588354 on Build GRCh38
rs547985564 Chr.5: 93588209-93588209 on Build GRCh38
rs771826396 Chr.5: 93588262-93588262 on Build GRCh38
rs898140239 Chr.5: 93588293-93588293 on Build GRCh38
rs761934434 Chr.5: 93588345-93588345 on Build GRCh38
rs754396202 Chr.5: 93588200-93588200 on Build GRCh38

View Full Product Details