Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41160625-41160850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP4-4
Entrez Gene ID
84616
Gene Name
keratin associated protein 4-4
Gene Aliases
KAP4.13, KAP4.4, KRTAP4-13, KRTAP4.13, KRTAP4.4
UniGene
Hs.307022
Gene Chromosome Location
Chr.17: 41159653-41160731 on Build GRCh38
Transcripts
NM_032524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5429434 c.9C>T p.N3N Chr.17: 41160683-41160683 on Build GRCh38
7352621 c.14G>T p.C5F Chr.17: 41160678-41160678 on Build GRCh38
379890 c.42C>T p.G14G Chr.17: 41160650-41160650 on Build GRCh38
2840085 c.63C>A p.C21* Chr.17: 41160629-41160629 on Build GRCh38
7079473 c.25G>A p.V9M Chr.17: 41160667-41160667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111435962 Chr.17: 41160678-41160678 on Build GRCh38
rs908424321 Chr.17: 41160790-41160790 on Build GRCh38
rs779913649 Chr.17: 41160741-41160741 on Build GRCh38
rs760910401 Chr.17: 41160675-41160675 on Build GRCh38
rs751178204 Chr.17: 41160708-41160708 on Build GRCh38
rs774099208 Chr.17: 41160686-41160687 on Build GRCh38
rs761864733 Chr.17: 41160663-41160663 on Build GRCh38
rs184690782 Chr.17: 41160762-41160762 on Build GRCh38
rs761143365 Chr.17: 41160696-41160696 on Build GRCh38
rs555128699 Chr.17: 41160781-41160781 on Build GRCh38
rs369803651 Chr.17: 41160684-41160684 on Build GRCh38
rs370704994 Chr.17: 41160680-41160680 on Build GRCh38
rs767618099 Chr.17: 41160667-41160667 on Build GRCh38
rs200769956 Chr.17: 41160689-41160689 on Build GRCh38
rs534376494 Chr.17: 41160677-41160677 on Build GRCh38
rs372425993 Chr.17: 41160669-41160669 on Build GRCh38
rs578174933 Chr.17: 41160804-41160804 on Build GRCh38
rs931099172 Chr.17: 41160764-41160764 on Build GRCh38
rs796615644 Chr.17: 41160782-41160782 on Build GRCh38
rs745701635 Chr.17: 41160719-41160719 on Build GRCh38
rs775466208 Chr.17: 41160654-41160654 on Build GRCh38
rs965396813 Chr.17: 41160734-41160734 on Build GRCh38
rs759093121 Chr.17: 41160687-41160688 on Build GRCh38
rs964942030 Chr.17: 41160670-41160670 on Build GRCh38
rs1056412268 Chr.17: 41160826-41160826 on Build GRCh38
rs990337892 Chr.17: 41160679-41160679 on Build GRCh38
rs780799736 Chr.17: 41160718-41160718 on Build GRCh38
rs556596107 Chr.17: 41160793-41160793 on Build GRCh38
rs769573071 Chr.17: 41160738-41160738 on Build GRCh38
rs757904227 Chr.17: 41160690-41160690 on Build GRCh38
rs756864683 Chr.17: 41160714-41160714 on Build GRCh38
rs893781997 Chr.17: 41160775-41160775 on Build GRCh38
rs912548618 Chr.17: 41160723-41160723 on Build GRCh38
rs376780195 Chr.17: 41160705-41160705 on Build GRCh38
rs975704850 Chr.17: 41160745-41160745 on Build GRCh38
rs765771593 Chr.17: 41160675-41160675 on Build GRCh38
rs781336341 Chr.17: 41160803-41160803 on Build GRCh38
rs1034620428 Chr.17: 41160645-41160645 on Build GRCh38
rs1001844040 Chr.17: 41160815-41160815 on Build GRCh38

View Full Product Details