Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1968762-1968991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF151
Entrez Gene ID
146310
Gene Name
ring finger protein 151
Gene Aliases
-
UniGene
Hs.99354
Gene Chromosome Location
Chr.16: 1966639-1968975 on Build GRCh38
Transcripts
NM_174903.4 XM_005255129.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376315 c.634C>A p.L212M Chr.16: 1968824-1968824 on Build GRCh38
1376314 c.632C>A p.A211D Chr.16: 1968822-1968822 on Build GRCh38
1323681 c.658_659insC p.E220fs*13 Chr.16: 1968848-1968849 on Build GRCh38
4883204 c.575C>G p.T192S Chr.16: 1968765-1968765 on Build GRCh38
4853708 c.601G>A p.E201K Chr.16: 1968791-1968791 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758481030 Chr.16: 1968787-1968787 on Build GRCh38
rs950574074 Chr.16: 1968845-1968845 on Build GRCh38
rs563164413 Chr.16: 1968915-1968915 on Build GRCh38
rs902640263 Chr.16: 1968885-1968885 on Build GRCh38
rs768709513 Chr.16: 1968945-1968945 on Build GRCh38
rs781468340 Chr.16: 1968839-1968839 on Build GRCh38
rs898510513 Chr.16: 1968964-1968964 on Build GRCh38
rs769950995 Chr.16: 1968850-1968850 on Build GRCh38
rs532140313 Chr.16: 1968821-1968821 on Build GRCh38
rs751528593 Chr.16: 1968784-1968784 on Build GRCh38
rs771948384 Chr.16: 1968968-1968968 on Build GRCh38
rs760351448 Chr.16: 1968884-1968884 on Build GRCh38
rs757771388 Chr.16: 1968924-1968924 on Build GRCh38
rs752102708 Chr.16: 1968923-1968923 on Build GRCh38
rs775619176 Chr.16: 1968852-1968852 on Build GRCh38
rs768725727 Chr.16: 1968860-1968860 on Build GRCh38
rs779583468 Chr.16: 1968820-1968820 on Build GRCh38
rs756524978 Chr.16: 1968938-1968938 on Build GRCh38
rs775278277 Chr.16: 1968875-1968875 on Build GRCh38
rs755786226 Chr.16: 1968811-1968811 on Build GRCh38
rs540124820 Chr.16: 1968927-1968927 on Build GRCh38
rs780480466 Chr.16: 1968837-1968837 on Build GRCh38
rs943159748 Chr.16: 1968866-1968866 on Build GRCh38
rs780181728 Chr.16: 1968939-1968939 on Build GRCh38
rs372562891 Chr.16: 1968881-1968881 on Build GRCh38
rs918676821 Chr.16: 1968849-1968849 on Build GRCh38
rs181399062 Chr.16: 1968876-1968876 on Build GRCh38
rs568231929 Chr.16: 1968946-1968946 on Build GRCh38
rs750076812 Chr.16: 1968794-1968794 on Build GRCh38
rs749524036 Chr.16: 1968943-1968943 on Build GRCh38
rs764069244 Chr.16: 1968783-1968783 on Build GRCh38
rs1030097326 Chr.16: 1968961-1968961 on Build GRCh38
rs761528355 Chr.16: 1968789-1968789 on Build GRCh38
rs774216491 Chr.16: 1968867-1968867 on Build GRCh38
rs371029957 Chr.16: 1968969-1968969 on Build GRCh38
rs1017336837 Chr.16: 1968941-1968941 on Build GRCh38
rs765896025 Chr.16: 1968889-1968889 on Build GRCh38
rs778386573 Chr.16: 1968830-1968830 on Build GRCh38
rs766203579 Chr.16: 1968971-1968971 on Build GRCh38
rs754473703 Chr.16: 1968898-1968898 on Build GRCh38
rs555573559 Chr.16: 1968892-1968892 on Build GRCh38
rs767230051 Chr.16: 1968791-1968791 on Build GRCh38
rs974516953 Chr.16: 1968843-1968843 on Build GRCh38
rs761796181 Chr.16: 1968874-1968874 on Build GRCh38
rs755537026 Chr.16: 1968891-1968892 on Build GRCh38
rs748043723 Chr.16: 1968949-1968949 on Build GRCh38
rs781589843 Chr.16: 1968926-1968926 on Build GRCh38
rs746098997 Chr.16: 1968840-1968840 on Build GRCh38
rs995141485 Chr.16: 1968966-1968966 on Build GRCh38
rs372986073 Chr.16: 1968854-1968854 on Build GRCh38
rs751820676 Chr.16: 1968843-1968845 on Build GRCh38
rs866535491 Chr.16: 1968864-1968864 on Build GRCh38
rs62617079 Chr.16: 1968911-1968911 on Build GRCh38

View Full Product Details