Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89666895-89667130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLIN1
Entrez Gene ID
5346
Gene Name
perilipin 1
Gene Aliases
FPLD4, PERI, PLIN
UniGene
Hs.103253
Gene Chromosome Location
Chr.15: 89664365-89679417 on Build GRCh38
Transcripts
NM_001145311.1 NM_002666.4 XM_005254934.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608816 c.1100T>C p.L367P Chr.15: 89667045-89667045 on Build GRCh38
3361581 c.1203C>T p.Y401Y Chr.15: 89666942-89666942 on Build GRCh38
40479 c.1134G>A p.G378G Chr.15: 89667011-89667011 on Build GRCh38
6015928 c.1209+9C>T p.? Chr.15: 89666927-89666927 on Build GRCh38
4128556 c.1099C>A p.L367I Chr.15: 89667046-89667046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370575940 Chr.15: 89666927-89666927 on Build GRCh38
rs756116966 Chr.15: 89666939-89666939 on Build GRCh38
rs140116505 Chr.15: 89667108-89667108 on Build GRCh38
rs151265993 Chr.15: 89666942-89666942 on Build GRCh38
rs751614860 Chr.15: 89667054-89667054 on Build GRCh38
rs952633761 Chr.15: 89666990-89666990 on Build GRCh38
rs367721916 Chr.15: 89666981-89666981 on Build GRCh38
rs752807112 Chr.15: 89666937-89666937 on Build GRCh38
rs752803680 Chr.15: 89667106-89667106 on Build GRCh38
rs764581818 Chr.15: 89666925-89666925 on Build GRCh38
rs746816190 Chr.15: 89667000-89667000 on Build GRCh38
rs751989453 Chr.15: 89666975-89666975 on Build GRCh38
rs769521575 Chr.15: 89667064-89667064 on Build GRCh38
rs771994170 Chr.15: 89667090-89667096 on Build GRCh38
rs199842965 Chr.15: 89667012-89667012 on Build GRCh38
rs770319244 Chr.15: 89667000-89667001 on Build GRCh38
rs774652319 Chr.15: 89667022-89667022 on Build GRCh38
rs1043065599 Chr.15: 89666946-89666946 on Build GRCh38
rs779527083 Chr.15: 89667069-89667069 on Build GRCh38
rs776564112 Chr.15: 89667003-89667003 on Build GRCh38
rs8179071 Chr.15: 89667102-89667102 on Build GRCh38
rs780230203 Chr.15: 89666984-89666984 on Build GRCh38
rs565394019 Chr.15: 89666918-89666918 on Build GRCh38
rs140394922 Chr.15: 89667009-89667009 on Build GRCh38
rs745508965 Chr.15: 89667032-89667033 on Build GRCh38
rs774850573 Chr.15: 89666962-89666962 on Build GRCh38
rs2304796 Chr.15: 89667026-89667026 on Build GRCh38
rs1056883929 Chr.15: 89667091-89667091 on Build GRCh38
rs764908291 Chr.15: 89666957-89666957 on Build GRCh38
rs146385147 Chr.15: 89667006-89667006 on Build GRCh38
rs763568117 Chr.15: 89666965-89666965 on Build GRCh38
rs141229584 Chr.15: 89667074-89667074 on Build GRCh38
rs754045975 Chr.15: 89666991-89666991 on Build GRCh38
rs1004121613 Chr.15: 89667031-89667031 on Build GRCh38
rs745631797 Chr.15: 89667063-89667063 on Build GRCh38
rs368908481 Chr.15: 89667076-89667076 on Build GRCh38
rs755810903 Chr.15: 89667111-89667111 on Build GRCh38
rs759124065 Chr.15: 89666951-89666951 on Build GRCh38
rs766470624 Chr.15: 89667039-89667039 on Build GRCh38
rs772535711 Chr.15: 89667085-89667085 on Build GRCh38
rs925542527 Chr.15: 89667107-89667107 on Build GRCh38
rs148207845 Chr.15: 89667049-89667049 on Build GRCh38
rs150128694 Chr.15: 89666966-89666966 on Build GRCh38
rs374661188 Chr.15: 89667029-89667029 on Build GRCh38
rs770728878 Chr.15: 89666944-89666944 on Build GRCh38
rs757249181 Chr.15: 89667055-89667055 on Build GRCh38
rs745574452 Chr.15: 89667004-89667004 on Build GRCh38
rs145599337 Chr.15: 89667019-89667019 on Build GRCh38
rs770784320 Chr.15: 89667001-89667001 on Build GRCh38
rs762352620 Chr.15: 89667082-89667082 on Build GRCh38
rs143876987 Chr.15: 89667040-89667040 on Build GRCh38
rs760698729 Chr.15: 89666977-89666977 on Build GRCh38
rs2304795 Chr.15: 89667032-89667032 on Build GRCh38
rs551819178 Chr.15: 89667087-89667087 on Build GRCh38
rs759801892 Chr.15: 89667101-89667101 on Build GRCh38
rs750268596 Chr.15: 89667109-89667109 on Build GRCh38
rs111761423 Chr.15: 89666935-89666935 on Build GRCh38
rs377142712 Chr.15: 89667094-89667094 on Build GRCh38
rs1051100757 Chr.15: 89666921-89666921 on Build GRCh38
rs754952710 Chr.15: 89667041-89667041 on Build GRCh38
rs779989440 Chr.15: 89666940-89666940 on Build GRCh38
rs8179072 Chr.15: 89666988-89666988 on Build GRCh38
rs777521631 Chr.15: 89666997-89666997 on Build GRCh38
rs371944125 Chr.15: 89667088-89667088 on Build GRCh38
rs189637126 Chr.15: 89666941-89666941 on Build GRCh38
rs750759829 Chr.15: 89667028-89667028 on Build GRCh38
rs750706792 Chr.15: 89666980-89666980 on Build GRCh38
rs757514141 Chr.15: 89666930-89666930 on Build GRCh38
rs2304797 Chr.15: 89666936-89666936 on Build GRCh38
rs373726332 Chr.15: 89666978-89666978 on Build GRCh38
rs140505022 Chr.15: 89666943-89666943 on Build GRCh38
rs776367759 Chr.15: 89666948-89666948 on Build GRCh38
rs781182993 Chr.15: 89667058-89667058 on Build GRCh38
rs748846601 Chr.15: 89667071-89667071 on Build GRCh38

View Full Product Details