Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48045587-48045860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT15
Entrez Gene ID
55270
Gene Name
nudix hydrolase 15
Gene Aliases
MTH2, NUDT15D
UniGene
Hs.144407
Gene Chromosome Location
Chr.13: 48037567-48052742 on Build GRCh38
Transcripts
NM_018283.3 NR_136687.1 NR_136688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947768 c.427G>T p.E143* Chr.13: 48045731-48045731 on Build GRCh38
5893745 c.481G>A p.G161R Chr.13: 48045785-48045785 on Build GRCh38
469479 c.450A>G p.K150K Chr.13: 48045754-48045754 on Build GRCh38
6587359 c.394G>C p.D132H Chr.13: 48045698-48045698 on Build GRCh38
947769 c.443C>A p.P148Q Chr.13: 48045747-48045747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036107704 Chr.13: 48045825-48045826 on Build GRCh38
rs776599118 Chr.13: 48045773-48045773 on Build GRCh38
rs778377158 Chr.13: 48045712-48045712 on Build GRCh38
rs751671087 Chr.13: 48045785-48045785 on Build GRCh38
rs961017463 Chr.13: 48045803-48045803 on Build GRCh38
rs376403083 Chr.13: 48045793-48045793 on Build GRCh38
rs764372517 Chr.13: 48045651-48045651 on Build GRCh38
rs943961420 Chr.13: 48045715-48045715 on Build GRCh38
rs772773911 Chr.13: 48045735-48045735 on Build GRCh38
rs753664856 Chr.13: 48045638-48045638 on Build GRCh38
rs749170586 Chr.13: 48045717-48045718 on Build GRCh38
rs773703261 Chr.13: 48045734-48045735 on Build GRCh38
rs147390019 Chr.13: 48045720-48045720 on Build GRCh38
rs778631332 Chr.13: 48045819-48045819 on Build GRCh38
rs757230015 Chr.13: 48045661-48045661 on Build GRCh38
rs768324690 Chr.13: 48045690-48045690 on Build GRCh38
rs747897258 Chr.13: 48045646-48045649 on Build GRCh38
rs775845592 Chr.13: 48045817-48045827 on Build GRCh38
rs139407085 Chr.13: 48045639-48045639 on Build GRCh38
rs369523087 Chr.13: 48045625-48045625 on Build GRCh38
rs755277002 Chr.13: 48045808-48045808 on Build GRCh38
rs749618301 Chr.13: 48045687-48045687 on Build GRCh38
rs781160995 Chr.13: 48045622-48045622 on Build GRCh38
rs370245521 Chr.13: 48045805-48045805 on Build GRCh38
rs760744843 Chr.13: 48045770-48045770 on Build GRCh38
rs781726752 Chr.13: 48045671-48045671 on Build GRCh38
rs764393847 Chr.13: 48045781-48045781 on Build GRCh38
rs373523703 Chr.13: 48045691-48045691 on Build GRCh38
rs751683170 Chr.13: 48045660-48045660 on Build GRCh38
rs772964646 Chr.13: 48045619-48045619 on Build GRCh38
rs200025929 Chr.13: 48045738-48045738 on Build GRCh38
rs746431481 Chr.13: 48045683-48045683 on Build GRCh38
rs777836864 Chr.13: 48045714-48045715 on Build GRCh38
rs116855232 Chr.13: 48045719-48045719 on Build GRCh38
rs139551410 Chr.13: 48045771-48045771 on Build GRCh38
rs780442234 Chr.13: 48045686-48045686 on Build GRCh38
rs766342196 Chr.13: 48045634-48045634 on Build GRCh38
rs771663636 Chr.13: 48045772-48045773 on Build GRCh38
rs750477947 Chr.13: 48045796-48045796 on Build GRCh38
rs770524351 Chr.13: 48045722-48045723 on Build GRCh38
rs1016725012 Chr.13: 48045727-48045727 on Build GRCh38
rs1026010382 Chr.13: 48045718-48045718 on Build GRCh38
rs372860978 Chr.13: 48045831-48045831 on Build GRCh38
rs200980982 Chr.13: 48045799-48045799 on Build GRCh38
rs747820731 Chr.13: 48045830-48045830 on Build GRCh38
rs756603433 Chr.13: 48045685-48045685 on Build GRCh38
rs375378556 Chr.13: 48045672-48045672 on Build GRCh38
rs745565963 Chr.13: 48045741-48045742 on Build GRCh38
rs769861583 Chr.13: 48045700-48045700 on Build GRCh38
rs762092824 Chr.13: 48045787-48045787 on Build GRCh38
rs61973267 Chr.13: 48045806-48045806 on Build GRCh38
rs763175763 Chr.13: 48045825-48045825 on Build GRCh38
rs142636030 Chr.13: 48045779-48045779 on Build GRCh38

View Full Product Details