Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8644688-8644915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL22
Entrez Gene ID
79091
Gene Name
methyltransferase like 22
Gene Aliases
C16orf68
UniGene
Hs.306380
Gene Chromosome Location
Chr.16: 8621640-8673565 on Build GRCh38
Transcripts
NM_024109.3 NR_134608.1 XM_005255570.3 XM_005255571.2 XM_011522657.2 XM_017023659.1 XM_017023660.1 XM_017023661.1 XM_017023662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3889111 c.1168C>T p.L390F Chr.16: 8644714-8644714 on Build GRCh38
3755168 c.1176A>G p.Q392Q Chr.16: 8644722-8644722 on Build GRCh38
5029774 c.1161C>T p.Y387Y Chr.16: 8644707-8644707 on Build GRCh38
7001006 c.1166G>TC p.R389fs*>17 Chr.16: 8644712-8644712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573232371 Chr.16: 8644859-8644859 on Build GRCh38
rs759378148 Chr.16: 8644750-8644750 on Build GRCh38
rs538812294 Chr.16: 8644829-8644829 on Build GRCh38
rs1730999 Chr.16: 8644784-8644784 on Build GRCh38
rs80169523 Chr.16: 8644843-8644843 on Build GRCh38
rs777938619 Chr.16: 8644841-8644841 on Build GRCh38
rs575875677 Chr.16: 8644842-8644842 on Build GRCh38
rs780763511 Chr.16: 8644716-8644716 on Build GRCh38
rs146697240 Chr.16: 8644882-8644882 on Build GRCh38
rs374919702 Chr.16: 8644728-8644728 on Build GRCh38
rs368969620 Chr.16: 8644774-8644774 on Build GRCh38
rs1044078115 Chr.16: 8644880-8644880 on Build GRCh38
rs373603242 Chr.16: 8644713-8644713 on Build GRCh38
rs1641067 Chr.16: 8644722-8644722 on Build GRCh38
rs1035744330 Chr.16: 8644787-8644787 on Build GRCh38
rs750802130 Chr.16: 8644775-8644775 on Build GRCh38
rs905200769 Chr.16: 8644886-8644886 on Build GRCh38
rs774162375 Chr.16: 8644740-8644740 on Build GRCh38
rs757855376 Chr.16: 8644747-8644747 on Build GRCh38
rs780640926 Chr.16: 8644782-8644782 on Build GRCh38
rs747080077 Chr.16: 8644720-8644720 on Build GRCh38
rs781360875 Chr.16: 8644788-8644788 on Build GRCh38
rs757506994 Chr.16: 8644710-8644710 on Build GRCh38
rs375819057 Chr.16: 8644714-8644714 on Build GRCh38
rs777918782 Chr.16: 8644798-8644798 on Build GRCh38
rs9933132 Chr.16: 8644765-8644765 on Build GRCh38
rs781210005 Chr.16: 8644733-8644733 on Build GRCh38
rs532300551 Chr.16: 8644708-8644708 on Build GRCh38
rs758898773 Chr.16: 8644776-8644776 on Build GRCh38
rs58228844 Chr.16: 8644768-8644768 on Build GRCh38
rs371062027 Chr.16: 8644712-8644712 on Build GRCh38
rs771986717 Chr.16: 8644751-8644751 on Build GRCh38
rs919457777 Chr.16: 8644891-8644891 on Build GRCh38
rs375713724 Chr.16: 8644752-8644752 on Build GRCh38
rs753534229 Chr.16: 8644709-8644709 on Build GRCh38
rs143114398 Chr.16: 8644794-8644794 on Build GRCh38
rs991875729 Chr.16: 8644819-8644819 on Build GRCh38
rs770588975 Chr.16: 8644737-8644737 on Build GRCh38
rs35262135 Chr.16: 8644840-8644840 on Build GRCh38
rs972323118 Chr.16: 8644878-8644878 on Build GRCh38
rs368307509 Chr.16: 8644735-8644735 on Build GRCh38
rs566128670 Chr.16: 8644766-8644766 on Build GRCh38
rs767486142 Chr.16: 8644707-8644707 on Build GRCh38
rs554704192 Chr.16: 8644856-8644856 on Build GRCh38
rs1730998 Chr.16: 8644851-8644851 on Build GRCh38
rs752121164 Chr.16: 8644783-8644783 on Build GRCh38

View Full Product Details