Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107167456-107167721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP62CL
Entrez Gene ID
54830
Gene Name
nucleoporin 62 C-terminal like
Gene Aliases
-
UniGene
Hs.163629
Gene Chromosome Location
Chr.X: 107123427-107206440 on Build GRCh38
Transcripts
NM_017681.2 NR_033676.1

SNP Detail

SNP ID Chromosome Location
rs1021473383 Chr.X: 107167596-107167596 on Build GRCh38
rs745957676 Chr.X: 107167625-107167625 on Build GRCh38
rs769770268 Chr.X: 107167640-107167640 on Build GRCh38
rs372408485 Chr.X: 107167609-107167609 on Build GRCh38
rs749546181 Chr.X: 107167665-107167665 on Build GRCh38
rs905526206 Chr.X: 107167519-107167519 on Build GRCh38
rs891753604 Chr.X: 107167520-107167520 on Build GRCh38
rs769923397 Chr.X: 107167627-107167628 on Build GRCh38
rs12845858 Chr.X: 107167574-107167574 on Build GRCh38
rs757701133 Chr.X: 107167691-107167691 on Build GRCh38
rs568817264 Chr.X: 107167695-107167695 on Build GRCh38
rs1011499101 Chr.X: 107167539-107167539 on Build GRCh38
rs16987290 Chr.X: 107167683-107167683 on Build GRCh38
rs376071089 Chr.X: 107167646-107167646 on Build GRCh38
rs775725736 Chr.X: 107167651-107167651 on Build GRCh38

View Full Product Details