Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233163486-233163747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
UniGene
Hs.262886
Gene Chromosome Location
Chr.2: 233059967-233207903 on Build GRCh38
Transcripts
NM_001017915.2 NM_005541.4 XM_017004004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3579204 c.1279C>T p.L427F Chr.2: 233163745-233163745 on Build GRCh38
3579203 c.1259A>C p.K420T Chr.2: 233163725-233163725 on Build GRCh38
4692599 c.1247delC p.P418fs*31 Chr.2: 233163713-233163713 on Build GRCh38
6090995 c.1263G>A p.K421K Chr.2: 233163729-233163729 on Build GRCh38
3049781 c.1246_1247insC p.P419fs*27 Chr.2: 233163712-233163713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887787035 Chr.2: 233163529-233163529 on Build GRCh38
rs764296244 Chr.2: 233163720-233163720 on Build GRCh38
rs969358739 Chr.2: 233163522-233163522 on Build GRCh38
rs373650820 Chr.2: 233163713-233163713 on Build GRCh38
rs564957593 Chr.2: 233163550-233163550 on Build GRCh38
rs751861631 Chr.2: 233163598-233163598 on Build GRCh38
rs985084517 Chr.2: 233163526-233163526 on Build GRCh38
rs749087989 Chr.2: 233163670-233163670 on Build GRCh38
rs575415857 Chr.2: 233163663-233163663 on Build GRCh38
rs754423970 Chr.2: 233163601-233163601 on Build GRCh38
rs1017922482 Chr.2: 233163609-233163609 on Build GRCh38
rs540249336 Chr.2: 233163551-233163551 on Build GRCh38
rs529998032 Chr.2: 233163540-233163540 on Build GRCh38
rs772238498 Chr.2: 233163681-233163681 on Build GRCh38
rs532648056 Chr.2: 233163608-233163608 on Build GRCh38
rs955268627 Chr.2: 233163590-233163590 on Build GRCh38
rs957020161 Chr.2: 233163626-233163626 on Build GRCh38
rs771607003 Chr.2: 233163538-233163538 on Build GRCh38
rs923423673 Chr.2: 233163539-233163539 on Build GRCh38
rs751255234 Chr.2: 233163721-233163721 on Build GRCh38
rs989854684 Chr.2: 233163628-233163628 on Build GRCh38
rs747161484 Chr.2: 233163704-233163704 on Build GRCh38
rs552483014 Chr.2: 233163666-233163666 on Build GRCh38
rs560388189 Chr.2: 233163586-233163586 on Build GRCh38
rs569340799 Chr.2: 233163711-233163711 on Build GRCh38
rs13385791 Chr.2: 233163528-233163528 on Build GRCh38
rs934558862 Chr.2: 233163667-233163667 on Build GRCh38
rs773495039 Chr.2: 233163687-233163687 on Build GRCh38
rs994386901 Chr.2: 233163552-233163552 on Build GRCh38
rs981989069 Chr.2: 233163672-233163672 on Build GRCh38
rs775892531 Chr.2: 233163718-233163718 on Build GRCh38
rs759837481 Chr.2: 233163712-233163713 on Build GRCh38
rs937383716 Chr.2: 233163717-233163717 on Build GRCh38
rs756013463 Chr.2: 233163515-233163515 on Build GRCh38
rs146216798 Chr.2: 233163530-233163530 on Build GRCh38
rs913457027 Chr.2: 233163648-233163648 on Build GRCh38
rs910257686 Chr.2: 233163566-233163566 on Build GRCh38
rs928838082 Chr.2: 233163714-233163714 on Build GRCh38

View Full Product Details