Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25585706-25585939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL39
Entrez Gene ID
54148
Gene Name
mitochondrial ribosomal protein L39
Gene Aliases
C21orf92, L39mt, MRP-L5, MRPL5, MSTP003, PRED22, PRED66, RPML5
UniGene
Hs.420696
Gene Chromosome Location
Chr.21: 25585656-25607858 on Build GRCh38
Transcripts
NM_017446.3 NM_080794.3 XM_011529651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162567 c.982C>T p.Q328* Chr.21: 25585742-25585742 on Build GRCh38
5858460 c.1000G>A p.E334K Chr.21: 25585724-25585724 on Build GRCh38
4703218 c.1001A>G p.E334G Chr.21: 25585723-25585723 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142513484 Chr.21: 25585733-25585733 on Build GRCh38
rs1038415083 Chr.21: 25585822-25585822 on Build GRCh38
rs998604526 Chr.21: 25585883-25585883 on Build GRCh38
rs957152535 Chr.21: 25585895-25585895 on Build GRCh38
rs1031390472 Chr.21: 25585884-25585884 on Build GRCh38
rs755603521 Chr.21: 25585782-25585782 on Build GRCh38
rs775872922 Chr.21: 25585794-25585794 on Build GRCh38
rs762478328 Chr.21: 25585801-25585801 on Build GRCh38
rs1029963396 Chr.21: 25585852-25585852 on Build GRCh38
rs543758167 Chr.21: 25585855-25585855 on Build GRCh38
rs367956972 Chr.21: 25585799-25585799 on Build GRCh38
rs200187839 Chr.21: 25585791-25585791 on Build GRCh38
rs747340967 Chr.21: 25585795-25585795 on Build GRCh38
rs577971565 Chr.21: 25585851-25585851 on Build GRCh38
rs754901467 Chr.21: 25585734-25585734 on Build GRCh38
rs769151461 Chr.21: 25585798-25585798 on Build GRCh38
rs772682916 Chr.21: 25585751-25585753 on Build GRCh38
rs887153424 Chr.21: 25585780-25585780 on Build GRCh38
rs899831044 Chr.21: 25585838-25585838 on Build GRCh38
rs1021346303 Chr.21: 25585815-25585815 on Build GRCh38
rs371994346 Chr.21: 25585784-25585784 on Build GRCh38
rs780993013 Chr.21: 25585745-25585745 on Build GRCh38
rs557999369 Chr.21: 25585809-25585809 on Build GRCh38
rs763332961 Chr.21: 25585864-25585864 on Build GRCh38
rs955249925 Chr.21: 25585875-25585875 on Build GRCh38
rs765380486 Chr.21: 25585806-25585806 on Build GRCh38
rs747632478 Chr.21: 25585768-25585768 on Build GRCh38
rs376277160 Chr.21: 25585788-25585788 on Build GRCh38
rs996802806 Chr.21: 25585849-25585849 on Build GRCh38

View Full Product Details