Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49622377-49622642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT2
Entrez Gene ID
4247
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Gene Aliases
CDG2A, CDGS2, GLCNACTII, GNT-II, GNT2
UniGene
Hs.93338
Gene Chromosome Location
Chr.14: 49620771-49623481 on Build GRCh38
Transcripts
NM_002408.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6899001 c.1154C>T p.T385I Chr.14: 49622422-49622422 on Build GRCh38
698480 c.1225G>C p.E409Q Chr.14: 49622493-49622493 on Build GRCh38
6874300 c.1210C>A p.Q404K Chr.14: 49622478-49622478 on Build GRCh38
1662756 c.1264A>C p.I422L Chr.14: 49622532-49622532 on Build GRCh38
955940 c.1231_1234delCTAA p.T412fs*8 Chr.14: 49622499-49622502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962841787 Chr.14: 49622502-49622502 on Build GRCh38
rs1006830352 Chr.14: 49622484-49622484 on Build GRCh38
rs746919419 Chr.14: 49622504-49622504 on Build GRCh38
rs762231765 Chr.14: 49622605-49622605 on Build GRCh38
rs759598046 Chr.14: 49622439-49622439 on Build GRCh38
rs761820792 Chr.14: 49622529-49622529 on Build GRCh38
rs759900257 Chr.14: 49622570-49622570 on Build GRCh38
rs781015590 Chr.14: 49622526-49622526 on Build GRCh38
rs373822224 Chr.14: 49622466-49622466 on Build GRCh38
rs563513161 Chr.14: 49622501-49622501 on Build GRCh38
rs950572878 Chr.14: 49622513-49622513 on Build GRCh38
rs34315441 Chr.14: 49622476-49622477 on Build GRCh38
rs199732373 Chr.14: 49622429-49622429 on Build GRCh38
rs764726477 Chr.14: 49622470-49622470 on Build GRCh38
rs370985289 Chr.14: 49622573-49622573 on Build GRCh38
rs759461580 Chr.14: 49622597-49622597 on Build GRCh38
rs3007039 Chr.14: 49622581-49622581 on Build GRCh38
rs754180253 Chr.14: 49622498-49622498 on Build GRCh38
rs772260019 Chr.14: 49622572-49622572 on Build GRCh38
rs61734676 Chr.14: 49622462-49622462 on Build GRCh38
rs769483436 Chr.14: 49622541-49622541 on Build GRCh38
rs767775001 Chr.14: 49622443-49622443 on Build GRCh38
rs774335811 Chr.14: 49622451-49622451 on Build GRCh38
rs144122741 Chr.14: 49622558-49622558 on Build GRCh38
rs140337098 Chr.14: 49622505-49622505 on Build GRCh38
rs747920154 Chr.14: 49622529-49622529 on Build GRCh38
rs979777755 Chr.14: 49622599-49622599 on Build GRCh38
rs199634745 Chr.14: 49622606-49622606 on Build GRCh38
rs200461712 Chr.14: 49622433-49622433 on Build GRCh38
rs776720499 Chr.14: 49622499-49622502 on Build GRCh38
rs146764245 Chr.14: 49622503-49622503 on Build GRCh38
rs757572881 Chr.14: 49622500-49622500 on Build GRCh38
rs1051276902 Chr.14: 49622431-49622431 on Build GRCh38
rs754301705 Chr.14: 49622598-49622598 on Build GRCh38

View Full Product Details