Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48459707-48459946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R21
Entrez Gene ID
129285
Gene Name
protein phosphatase 1 regulatory subunit 21
Gene Aliases
CCDC128, KLRAQ1
UniGene
Hs.654619
Gene Chromosome Location
Chr.2: 48440769-48515392 on Build GRCh38
Transcripts
NM_001135629.2 NM_001193475.1 NM_152994.4 NR_024188.2 XM_017003301.1 XM_017003302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6304146 c.488G>A p.R163Q Chr.2: 48459866-48459866 on Build GRCh38
721715 c.486C>T p.T162T Chr.2: 48459864-48459864 on Build GRCh38
6251078 c.513G>T p.K171N Chr.2: 48459891-48459891 on Build GRCh38
6286831 c.496A>G p.M166V Chr.2: 48459874-48459874 on Build GRCh38
6251077 c.513G>T p.K171N Chr.2: 48459891-48459891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777231699 Chr.2: 48459825-48459825 on Build GRCh38
rs751027468 Chr.2: 48459897-48459897 on Build GRCh38
rs769744843 Chr.2: 48459903-48459903 on Build GRCh38
rs766687094 Chr.2: 48459872-48459872 on Build GRCh38
rs1017221402 Chr.2: 48459884-48459884 on Build GRCh38
rs971083463 Chr.2: 48459800-48459800 on Build GRCh38
rs769656049 Chr.2: 48459838-48459838 on Build GRCh38
rs749290174 Chr.2: 48459908-48459908 on Build GRCh38
rs200936594 Chr.2: 48459806-48459806 on Build GRCh38
rs750744374 Chr.2: 48459775-48459775 on Build GRCh38
rs770476503 Chr.2: 48459832-48459832 on Build GRCh38
rs780723061 Chr.2: 48459902-48459902 on Build GRCh38
rs370378310 Chr.2: 48459874-48459874 on Build GRCh38
rs767778479 Chr.2: 48459906-48459907 on Build GRCh38
rs764223391 Chr.2: 48459743-48459743 on Build GRCh38
rs756430637 Chr.2: 48459796-48459796 on Build GRCh38
rs922682647 Chr.2: 48459883-48459883 on Build GRCh38
rs536120802 Chr.2: 48459865-48459865 on Build GRCh38
rs557336595 Chr.2: 48459841-48459841 on Build GRCh38
rs200719129 Chr.2: 48459763-48459763 on Build GRCh38
rs751840727 Chr.2: 48459746-48459746 on Build GRCh38
rs757931817 Chr.2: 48459809-48459809 on Build GRCh38
rs754749964 Chr.2: 48459866-48459866 on Build GRCh38
rs756719736 Chr.2: 48459898-48459898 on Build GRCh38
rs762946506 Chr.2: 48459853-48459853 on Build GRCh38
rs376812012 Chr.2: 48459834-48459834 on Build GRCh38
rs767633163 Chr.2: 48459859-48459859 on Build GRCh38
rs146977827 Chr.2: 48459753-48459753 on Build GRCh38
rs757392695 Chr.2: 48459752-48459752 on Build GRCh38
rs781108104 Chr.2: 48459833-48459833 on Build GRCh38
rs964895733 Chr.2: 48459869-48459869 on Build GRCh38
rs768899408 Chr.2: 48459855-48459855 on Build GRCh38
rs777321546 Chr.2: 48459888-48459888 on Build GRCh38
rs142054593 Chr.2: 48459856-48459856 on Build GRCh38
rs762157853 Chr.2: 48459857-48459857 on Build GRCh38
rs754292034 Chr.2: 48459808-48459808 on Build GRCh38
rs753871601 Chr.2: 48459814-48459814 on Build GRCh38
rs746722848 Chr.2: 48459831-48459831 on Build GRCh38
rs763140258 Chr.2: 48459741-48459741 on Build GRCh38
rs998498095 Chr.2: 48459805-48459805 on Build GRCh38

View Full Product Details