Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70530690-70530963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SF3B3
Entrez Gene ID
23450
Gene Name
splicing factor 3b subunit 3
Gene Aliases
RSE1, SAP130, SF3b130, STAF130
UniGene
Hs.514435
Gene Chromosome Location
Chr.16: 70523788-70577668 on Build GRCh38
Transcripts
NM_012426.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5740716 c.431T>C p.L144S Chr.16: 70530778-70530778 on Build GRCh38
1379316 c.419T>C p.L140S Chr.16: 70530766-70530766 on Build GRCh38
7150618 c.432G>T p.L144F Chr.16: 70530779-70530779 on Build GRCh38
4499276 c.538C>T p.P180S Chr.16: 70530885-70530885 on Build GRCh38
973036 c.565T>C p.Y189H Chr.16: 70530912-70530912 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009969585 Chr.16: 70530736-70530736 on Build GRCh38
rs763495044 Chr.16: 70530898-70530898 on Build GRCh38
rs750260538 Chr.16: 70530737-70530737 on Build GRCh38
rs960874459 Chr.16: 70530887-70530887 on Build GRCh38
rs1042949152 Chr.16: 70530934-70530934 on Build GRCh38
rs1022328014 Chr.16: 70530763-70530763 on Build GRCh38
rs1039505233 Chr.16: 70530806-70530806 on Build GRCh38
rs755997535 Chr.16: 70530741-70530741 on Build GRCh38
rs992347665 Chr.16: 70530908-70530908 on Build GRCh38
rs780093986 Chr.16: 70530742-70530742 on Build GRCh38
rs758542724 Chr.16: 70530754-70530754 on Build GRCh38
rs768243788 Chr.16: 70530941-70530941 on Build GRCh38
rs747182921 Chr.16: 70530819-70530819 on Build GRCh38
rs1016243949 Chr.16: 70530928-70530928 on Build GRCh38
rs947322967 Chr.16: 70530853-70530853 on Build GRCh38
rs376818809 Chr.16: 70530818-70530818 on Build GRCh38
rs752695674 Chr.16: 70530747-70530747 on Build GRCh38
rs776115986 Chr.16: 70530936-70530936 on Build GRCh38
rs200788579 Chr.16: 70530719-70530719 on Build GRCh38
rs767376375 Chr.16: 70530722-70530722 on Build GRCh38
rs781467882 Chr.16: 70530878-70530878 on Build GRCh38
rs770188490 Chr.16: 70530929-70530929 on Build GRCh38
rs771154873 Chr.16: 70530872-70530872 on Build GRCh38
rs1002247850 Chr.16: 70530854-70530854 on Build GRCh38

View Full Product Details