Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33477929-33478202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEPD
Entrez Gene ID
5184
Gene Name
peptidase D
Gene Aliases
PROLIDASE
UniGene
Hs.36473
Gene Chromosome Location
Chr.19: 33386949-33521893 on Build GRCh38
Transcripts
NM_000285.3 NM_001166056.1 NM_001166057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
262901 c.507C>T p.F169F Chr.19: 33478087-33478087 on Build GRCh38
5187400 c.541G>A p.V181I Chr.19: 33478053-33478053 on Build GRCh38
4076796 c.540C>T p.I180I Chr.19: 33478054-33478054 on Build GRCh38
6049527 c.504-9G>A p.? Chr.19: 33478099-33478099 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775561400 Chr.19: 33478126-33478126 on Build GRCh38
rs765256652 Chr.19: 33478056-33478056 on Build GRCh38
rs774305529 Chr.19: 33478041-33478041 on Build GRCh38
rs377714630 Chr.19: 33478087-33478087 on Build GRCh38
rs758711931 Chr.19: 33478002-33478002 on Build GRCh38
rs376552339 Chr.19: 33478135-33478135 on Build GRCh38
rs901716483 Chr.19: 33478164-33478164 on Build GRCh38
rs368784737 Chr.19: 33478083-33478083 on Build GRCh38
rs564331867 Chr.19: 33478145-33478145 on Build GRCh38
rs897864057 Chr.19: 33477985-33477985 on Build GRCh38
rs116191310 Chr.19: 33477959-33477959 on Build GRCh38
rs981571305 Chr.19: 33478117-33478117 on Build GRCh38
rs764384475 Chr.19: 33478079-33478079 on Build GRCh38
rs758357853 Chr.19: 33478086-33478086 on Build GRCh38
rs753194722 Chr.19: 33477999-33477999 on Build GRCh38
rs753270602 Chr.19: 33477994-33477994 on Build GRCh38
rs375333595 Chr.19: 33478031-33478031 on Build GRCh38
rs780630338 Chr.19: 33478011-33478011 on Build GRCh38
rs746106960 Chr.19: 33478114-33478114 on Build GRCh38
rs1046222558 Chr.19: 33477963-33477963 on Build GRCh38
rs751146313 Chr.19: 33478004-33478004 on Build GRCh38
rs75766592 Chr.19: 33478099-33478099 on Build GRCh38
rs1015860025 Chr.19: 33478154-33478154 on Build GRCh38
rs781428367 Chr.19: 33478104-33478104 on Build GRCh38
rs754956964 Chr.19: 33478093-33478093 on Build GRCh38
rs369357394 Chr.19: 33478037-33478037 on Build GRCh38
rs780762135 Chr.19: 33478018-33478018 on Build GRCh38
rs565998298 Chr.19: 33477970-33477970 on Build GRCh38
rs145109685 Chr.19: 33478137-33478137 on Build GRCh38
rs368437163 Chr.19: 33477977-33477977 on Build GRCh38
rs534952816 Chr.19: 33478005-33478005 on Build GRCh38
rs769143183 Chr.19: 33478128-33478128 on Build GRCh38
rs746885456 Chr.19: 33478127-33478127 on Build GRCh38
rs993885514 Chr.19: 33477986-33477986 on Build GRCh38
rs745374508 Chr.19: 33478019-33478019 on Build GRCh38
rs776024032 Chr.19: 33478058-33478058 on Build GRCh38
rs372629704 Chr.19: 33478053-33478053 on Build GRCh38
rs376361050 Chr.19: 33478060-33478060 on Build GRCh38
rs950501608 Chr.19: 33478143-33478143 on Build GRCh38
rs542228812 Chr.19: 33478091-33478091 on Build GRCh38
rs549565751 Chr.19: 33478174-33478174 on Build GRCh38
rs201605550 Chr.19: 33478100-33478100 on Build GRCh38
rs372377642 Chr.19: 33478003-33478003 on Build GRCh38
rs762140257 Chr.19: 33478133-33478133 on Build GRCh38
rs769843610 Chr.19: 33478118-33478118 on Build GRCh38
rs557965371 Chr.19: 33478026-33478026 on Build GRCh38
rs753775083 Chr.19: 33478092-33478092 on Build GRCh38
rs774874484 Chr.19: 33478129-33478129 on Build GRCh38
rs61748998 Chr.19: 33478085-33478085 on Build GRCh38
rs780015592 Chr.19: 33478030-33478030 on Build GRCh38
rs759640890 Chr.19: 33478054-33478054 on Build GRCh38
rs146834287 Chr.19: 33478167-33478167 on Build GRCh38
rs556625829 Chr.19: 33478046-33478046 on Build GRCh38
rs370589638 Chr.19: 33478136-33478136 on Build GRCh38
rs1021121551 Chr.19: 33478077-33478077 on Build GRCh38
rs766117480 Chr.19: 33478141-33478141 on Build GRCh38
rs370887337 Chr.19: 33478096-33478096 on Build GRCh38

View Full Product Details