Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197003774-197004029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MELTF
Entrez Gene ID
4241
Gene Name
melanotransferrin
Gene Aliases
CD228, MAP97, MFI2, MTF1, MTf
UniGene
Hs.184727
Gene Chromosome Location
Chr.3: 197001740-197029816 on Build GRCh38
Transcripts
NM_005929.5 XM_006713643.3 XM_011512850.2 NR_038285.1

Gene Symbol
MFI2-AS1
Entrez Gene ID
100507057
Gene Name
MFI2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.666451
Gene Chromosome Location
Chr.3: 197002906-197004744 on Build GRCh38
Transcripts
NM_005929.5 XM_006713643.3 XM_011512850.2 NR_038285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370217 c.2075G>A p.W692* Chr.3: 197003963-197003963 on Build GRCh38
3847169 c.2117C>T p.S706F Chr.3: 197003921-197003921 on Build GRCh38
6249943 c.2099C>A p.A700E Chr.3: 197003939-197003939 on Build GRCh38
1422059 c.2112G>A p.M704I Chr.3: 197003926-197003926 on Build GRCh38
1308988 c.2027C>T p.A676V Chr.3: 197004011-197004011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920734833 Chr.3: 197003809-197003809 on Build GRCh38
rs761850068 Chr.3: 197003928-197003928 on Build GRCh38
rs200400304 Chr.3: 197003896-197003896 on Build GRCh38
rs1008902303 Chr.3: 197003879-197003879 on Build GRCh38
rs751296070 Chr.3: 197003947-197003947 on Build GRCh38
rs776410241 Chr.3: 197003919-197003919 on Build GRCh38
rs778253146 Chr.3: 197003955-197003955 on Build GRCh38
rs764588336 Chr.3: 197003921-197003923 on Build GRCh38
rs959631683 Chr.3: 197004002-197004002 on Build GRCh38
rs775782894 Chr.3: 197003906-197003906 on Build GRCh38
rs761808524 Chr.3: 197003917-197003917 on Build GRCh38
rs911079086 Chr.3: 197003802-197003802 on Build GRCh38
rs1000382853 Chr.3: 197003934-197003934 on Build GRCh38
rs938209319 Chr.3: 197003791-197003791 on Build GRCh38
rs372763159 Chr.3: 197003935-197003935 on Build GRCh38
rs201574477 Chr.3: 197003989-197003989 on Build GRCh38
rs749868484 Chr.3: 197003891-197003891 on Build GRCh38
rs754180944 Chr.3: 197003957-197003957 on Build GRCh38
rs747297468 Chr.3: 197003988-197003988 on Build GRCh38
rs962585809 Chr.3: 197003818-197003818 on Build GRCh38
rs1044537523 Chr.3: 197003923-197003923 on Build GRCh38
rs574937919 Chr.3: 197003892-197003892 on Build GRCh38
rs769651004 Chr.3: 197004004-197004004 on Build GRCh38
rs1037814200 Chr.3: 197003803-197003803 on Build GRCh38
rs758007124 Chr.3: 197003865-197003865 on Build GRCh38
rs768552718 Chr.3: 197003909-197003909 on Build GRCh38
rs879000036 Chr.3: 197003800-197003800 on Build GRCh38
rs199526102 Chr.3: 197003967-197003967 on Build GRCh38
rs375143966 Chr.3: 197003993-197003993 on Build GRCh38
rs375645349 Chr.3: 197003900-197003900 on Build GRCh38
rs148977686 Chr.3: 197003946-197003946 on Build GRCh38
rs113908132 Chr.3: 197003823-197003823 on Build GRCh38
rs974033008 Chr.3: 197004006-197004006 on Build GRCh38
rs779705590 Chr.3: 197003868-197003868 on Build GRCh38
rs570885404 Chr.3: 197003837-197003837 on Build GRCh38
rs764776254 Chr.3: 197003851-197003851 on Build GRCh38
rs1033915213 Chr.3: 197003992-197003992 on Build GRCh38
rs41292039 Chr.3: 197003888-197003888 on Build GRCh38
rs369001558 Chr.3: 197003905-197003905 on Build GRCh38
rs947832091 Chr.3: 197003916-197003916 on Build GRCh38
rs141948372 Chr.3: 197004007-197004007 on Build GRCh38
rs201948273 Chr.3: 197003950-197003950 on Build GRCh38
rs761626675 Chr.3: 197003997-197003997 on Build GRCh38
rs778142017 Chr.3: 197003890-197003890 on Build GRCh38
rs370646388 Chr.3: 197003968-197003968 on Build GRCh38
rs143354088 Chr.3: 197003942-197003942 on Build GRCh38
rs752949147 Chr.3: 197003869-197003869 on Build GRCh38
rs779573004 Chr.3: 197003958-197003958 on Build GRCh38
rs538311719 Chr.3: 197003860-197003860 on Build GRCh38

View Full Product Details