Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108497302-108497559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA8
Entrez Gene ID
50640
Gene Name
patatin like phospholipase domain containing 8
Gene Aliases
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma
UniGene
Hs.617340
Gene Chromosome Location
Chr.7: 108470417-108528161 on Build GRCh38
Transcripts
NM_001256007.2 NM_001256008.2 NM_001256009.2 NM_001256010.2 NM_001256011.2 NM_015723.4 XM_005250396.4 XM_011516274.2 XM_011516275.2 XM_017012261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6108045 c.1409A>C p.K470T Chr.7: 108497527-108497527 on Build GRCh38
6751573 c.1443T>C p.G481G Chr.7: 108497493-108497493 on Build GRCh38
3922482 c.1380C>T p.T460T Chr.7: 108497556-108497556 on Build GRCh38
3877167 c.1381C>A p.L461I Chr.7: 108497555-108497555 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745944132 Chr.7: 108497442-108497442 on Build GRCh38
rs773651908 Chr.7: 108497502-108497502 on Build GRCh38
rs1052878336 Chr.7: 108497330-108497330 on Build GRCh38
rs766929591 Chr.7: 108497338-108497338 on Build GRCh38
rs781054305 Chr.7: 108497439-108497439 on Build GRCh38
rs1042449300 Chr.7: 108497352-108497352 on Build GRCh38
rs537104627 Chr.7: 108497431-108497431 on Build GRCh38
rs556152762 Chr.7: 108497488-108497488 on Build GRCh38
rs568217039 Chr.7: 108497429-108497429 on Build GRCh38
rs760094418 Chr.7: 108497500-108497500 on Build GRCh38
rs935574330 Chr.7: 108497321-108497321 on Build GRCh38
rs902154854 Chr.7: 108497374-108497374 on Build GRCh38
rs946283043 Chr.7: 108497345-108497348 on Build GRCh38
rs774873730 Chr.7: 108497473-108497473 on Build GRCh38
rs996044493 Chr.7: 108497418-108497418 on Build GRCh38
rs548148756 Chr.7: 108497393-108497393 on Build GRCh38
rs998416170 Chr.7: 108497461-108497461 on Build GRCh38
rs372465092 Chr.7: 108497469-108497469 on Build GRCh38
rs769811223 Chr.7: 108497456-108497456 on Build GRCh38
rs900234446 Chr.7: 108497416-108497416 on Build GRCh38
rs771547993 Chr.7: 108497469-108497471 on Build GRCh38
rs1039762222 Chr.7: 108497427-108497427 on Build GRCh38
rs562455761 Chr.7: 108497415-108497415 on Build GRCh38
rs1032121550 Chr.7: 108497405-108497405 on Build GRCh38
rs1015353716 Chr.7: 108497465-108497465 on Build GRCh38
rs762343631 Chr.7: 108497493-108497493 on Build GRCh38
rs772615573 Chr.7: 108497480-108497481 on Build GRCh38
rs777332505 Chr.7: 108497422-108497422 on Build GRCh38
rs369340269 Chr.7: 108497385-108497385 on Build GRCh38
rs368765287 Chr.7: 108497472-108497472 on Build GRCh38

View Full Product Details