Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5211367-5211634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
UniGene
Hs.744928
Gene Chromosome Location
Chr.19: 5205503-5340803 on Build GRCh38
Transcripts
NM_002850.3 NM_130853.2 NM_130854.2 NM_130855.2 XM_005259600.2 XM_005259606.2 XM_005259607.2 XM_005259609.1 XM_005259610.1 XM_011528157.2 XM_011528158.2 XM_017027065.1 XM_017027066.1 XM_017027067.1 XM_017027068.1 XM_017027069.1 XM_017027070.1 XM_017027071.1 XM_017027072.1 XM_017027073.1 XM_017027074.1 XM_017027075.1 XM_017027076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275455 c.5237+8C>A p.? Chr.19: 5211582-5211582 on Build GRCh38
7275454 c.5234+8C>A p.? Chr.19: 5211582-5211582 on Build GRCh38
6936581 c.5198C>T p.S1733F Chr.19: 5211626-5211626 on Build GRCh38
6936580 c.5201C>T p.S1734F Chr.19: 5211626-5211626 on Build GRCh38
6951831 c.5213C>T p.A1738V Chr.19: 5211611-5211611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139609579 Chr.19: 5211528-5211528 on Build GRCh38
rs776788648 Chr.19: 5211563-5211564 on Build GRCh38
rs759140443 Chr.19: 5211509-5211510 on Build GRCh38
rs756263229 Chr.19: 5211595-5211595 on Build GRCh38
rs935650525 Chr.19: 5211473-5211473 on Build GRCh38
rs992510803 Chr.19: 5211579-5211579 on Build GRCh38
rs567493734 Chr.19: 5211598-5211598 on Build GRCh38
rs111588397 Chr.19: 5211532-5211532 on Build GRCh38
rs41276864 Chr.19: 5211601-5211601 on Build GRCh38
rs968440753 Chr.19: 5211534-5211534 on Build GRCh38
rs751933863 Chr.19: 5211568-5211568 on Build GRCh38
rs545521843 Chr.19: 5211421-5211421 on Build GRCh38
rs776672620 Chr.19: 5211549-5211549 on Build GRCh38
rs1018257048 Chr.19: 5211501-5211501 on Build GRCh38
rs1006958204 Chr.19: 5211455-5211455 on Build GRCh38
rs897080223 Chr.19: 5211426-5211426 on Build GRCh38
rs766789243 Chr.19: 5211567-5211567 on Build GRCh38
rs981410187 Chr.19: 5211462-5211462 on Build GRCh38
rs762510667 Chr.19: 5211566-5211566 on Build GRCh38
rs148510043 Chr.19: 5211428-5211428 on Build GRCh38
rs969588823 Chr.19: 5211432-5211432 on Build GRCh38
rs761412526 Chr.19: 5211553-5211553 on Build GRCh38
rs926972095 Chr.19: 5211572-5211572 on Build GRCh38
rs887746679 Chr.19: 5211418-5211418 on Build GRCh38
rs753096321 Chr.19: 5211585-5211585 on Build GRCh38
rs925569058 Chr.19: 5211467-5211467 on Build GRCh38
rs757537399 Chr.19: 5211602-5211602 on Build GRCh38
rs768042925 Chr.19: 5211581-5211581 on Build GRCh38
rs764635172 Chr.19: 5211556-5211556 on Build GRCh38
rs151021716 Chr.19: 5211506-5211506 on Build GRCh38
rs528053428 Chr.19: 5211531-5211531 on Build GRCh38
rs538427330 Chr.19: 5211412-5211412 on Build GRCh38
rs543649800 Chr.19: 5211429-5211429 on Build GRCh38
rs998439600 Chr.19: 5211521-5211521 on Build GRCh38
rs115796999 Chr.19: 5211613-5211613 on Build GRCh38
rs146317619 Chr.19: 5211422-5211422 on Build GRCh38
rs55858361 Chr.19: 5211557-5211557 on Build GRCh38
rs536709073 Chr.19: 5211548-5211548 on Build GRCh38
rs142784771 Chr.19: 5211434-5211434 on Build GRCh38
rs202165573 Chr.19: 5211543-5211543 on Build GRCh38
rs1141373 Chr.19: 5211592-5211592 on Build GRCh38
rs888502366 Chr.19: 5211430-5211430 on Build GRCh38
rs192294702 Chr.19: 5211527-5211527 on Build GRCh38
rs1031636298 Chr.19: 5211522-5211522 on Build GRCh38
rs759827526 Chr.19: 5211576-5211576 on Build GRCh38

View Full Product Details