Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75265821-75266064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS7
Entrez Gene ID
51081
Gene Name
mitochondrial ribosomal protein S7
Gene Aliases
MRP-S, MRP-S7, RP-S7, RPMS7, S7mt, bMRP27a
UniGene
Hs.71787
Gene Chromosome Location
Chr.17: 75261668-75266376 on Build GRCh38
Transcripts
NM_015971.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6082068 c.635C>T p.A212V Chr.17: 75265829-75265829 on Build GRCh38
6715530 c.700C>T p.R234C Chr.17: 75265894-75265894 on Build GRCh38
6842212 c.653C>A p.P218H Chr.17: 75265847-75265847 on Build GRCh38
5621287 c.637T>C p.F213L Chr.17: 75265831-75265831 on Build GRCh38
437372 c.666G>A p.R222R Chr.17: 75265860-75265860 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778531303 Chr.17: 75265913-75265913 on Build GRCh38
rs755150022 Chr.17: 75265894-75265894 on Build GRCh38
rs773407145 Chr.17: 75265869-75265869 on Build GRCh38
rs1018920039 Chr.17: 75266009-75266009 on Build GRCh38
rs753034365 Chr.17: 75265898-75265898 on Build GRCh38
rs1045564385 Chr.17: 75265903-75265903 on Build GRCh38
rs768429856 Chr.17: 75265948-75265948 on Build GRCh38
rs373944549 Chr.17: 75265849-75265849 on Build GRCh38
rs767905629 Chr.17: 75265895-75265895 on Build GRCh38
rs559829368 Chr.17: 75265981-75265981 on Build GRCh38
rs745556197 Chr.17: 75265920-75265920 on Build GRCh38
rs148321039 Chr.17: 75265848-75265848 on Build GRCh38
rs201977840 Chr.17: 75265954-75265954 on Build GRCh38
rs774378361 Chr.17: 75265964-75265964 on Build GRCh38
rs1043624833 Chr.17: 75265982-75265982 on Build GRCh38
rs762944168 Chr.17: 75265880-75265880 on Build GRCh38
rs746777155 Chr.17: 75265943-75265943 on Build GRCh38
rs778150192 Chr.17: 75265902-75265902 on Build GRCh38
rs757678398 Chr.17: 75265972-75265972 on Build GRCh38
rs115093728 Chr.17: 75265882-75265882 on Build GRCh38
rs775485351 Chr.17: 75265942-75265942 on Build GRCh38
rs146355809 Chr.17: 75265905-75265905 on Build GRCh38
rs892290289 Chr.17: 75265960-75265960 on Build GRCh38
rs767709317 Chr.17: 75265974-75265974 on Build GRCh38
rs764521589 Chr.17: 75265857-75265866 on Build GRCh38
rs751801208 Chr.17: 75265889-75265889 on Build GRCh38
rs528940496 Chr.17: 75265972-75265972 on Build GRCh38
rs756743610 Chr.17: 75265912-75265912 on Build GRCh38
rs763925773 Chr.17: 75265857-75265857 on Build GRCh38
rs150992484 Chr.17: 75266005-75266005 on Build GRCh38
rs965660517 Chr.17: 75266020-75266020 on Build GRCh38
rs771828845 Chr.17: 75265922-75265922 on Build GRCh38
rs759799301 Chr.17: 75265967-75265967 on Build GRCh38
rs759219286 Chr.17: 75265843-75265843 on Build GRCh38
rs902559649 Chr.17: 75265910-75265910 on Build GRCh38
rs116788769 Chr.17: 75265901-75265901 on Build GRCh38
rs200732026 Chr.17: 75265955-75265955 on Build GRCh38
rs372684480 Chr.17: 75266003-75266003 on Build GRCh38
rs754218722 Chr.17: 75265922-75265923 on Build GRCh38

View Full Product Details