Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57318385-57318658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRIM2
Entrez Gene ID
5558
Gene Name
primase (DNA) subunit 2
Gene Aliases
PRIM2A, p58
UniGene
Hs.654580
Gene Chromosome Location
Chr.6: 57314805-57646852 on Build GRCh38
Transcripts
NM_000947.4 NM_001282487.1 NM_001282488.1 XM_017011004.1 XM_017011005.1 XM_017011006.1 XM_017011007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312335 c.33G>A p.L11L Chr.6: 57318478-57318478 on Build GRCh38
1221984 c.154+1G>T p.? Chr.6: 57318600-57318600 on Build GRCh38
263097 c.121G>T p.E41* Chr.6: 57318566-57318566 on Build GRCh38
7416159 c.75C>A p.C25* Chr.6: 57318520-57318520 on Build GRCh38
5658890 c.79C>T p.Q27* Chr.6: 57318524-57318524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770994019 Chr.6: 57318550-57318550 on Build GRCh38
rs375453553 Chr.6: 57318609-57318609 on Build GRCh38
rs754524659 Chr.6: 57318491-57318491 on Build GRCh38
rs768638282 Chr.6: 57318494-57318494 on Build GRCh38
rs749727827 Chr.6: 57318413-57318413 on Build GRCh38
rs749559399 Chr.6: 57318526-57318526 on Build GRCh38
rs892930974 Chr.6: 57318598-57318598 on Build GRCh38
rs767576328 Chr.6: 57318571-57318571 on Build GRCh38
rs757938591 Chr.6: 57318481-57318481 on Build GRCh38
rs373417300 Chr.6: 57318517-57318517 on Build GRCh38
rs369094479 Chr.6: 57318412-57318412 on Build GRCh38
rs769679198 Chr.6: 57318512-57318512 on Build GRCh38
rs923381110 Chr.6: 57318438-57318438 on Build GRCh38
rs764330737 Chr.6: 57318445-57318445 on Build GRCh38
rs750138475 Chr.6: 57318462-57318462 on Build GRCh38
rs762467299 Chr.6: 57318430-57318433 on Build GRCh38
rs1055559616 Chr.6: 57318507-57318507 on Build GRCh38
rs1046942040 Chr.6: 57318560-57318560 on Build GRCh38
rs756565450 Chr.6: 57318433-57318433 on Build GRCh38
rs933782458 Chr.6: 57318502-57318502 on Build GRCh38
rs746450660 Chr.6: 57318489-57318489 on Build GRCh38
rs183241138 Chr.6: 57318554-57318554 on Build GRCh38
rs558224198 Chr.6: 57318425-57318425 on Build GRCh38
rs779741028 Chr.6: 57318487-57318487 on Build GRCh38
rs570710852 Chr.6: 57318501-57318501 on Build GRCh38
rs530852393 Chr.6: 57318422-57318422 on Build GRCh38
rs780944762 Chr.6: 57318586-57318586 on Build GRCh38
rs759666690 Chr.6: 57318557-57318557 on Build GRCh38

View Full Product Details