Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29485656-29485917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEFH
Entrez Gene ID
4744
Gene Name
neurofilament heavy polypeptide
Gene Aliases
CMT2CC, NFH
UniGene
Hs.198760
Gene Chromosome Location
Chr.22: 29480192-29491290 on Build GRCh38
Transcripts
NM_021076.3 XM_011530200.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2936486 c.1138G>A p.A380T Chr.22: 29485777-29485777 on Build GRCh38
6162005 c.1134G>C p.M378I Chr.22: 29485773-29485773 on Build GRCh38
5366682 c.1137C>T p.A379A Chr.22: 29485776-29485776 on Build GRCh38
3553169 c.1165C>A p.L389I Chr.22: 29485804-29485804 on Build GRCh38
5456947 c.1162C>A p.L388M Chr.22: 29485801-29485801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376113957 Chr.22: 29485876-29485876 on Build GRCh38
rs165734 Chr.22: 29485839-29485839 on Build GRCh38
rs752243905 Chr.22: 29485820-29485820 on Build GRCh38
rs757021413 Chr.22: 29485838-29485838 on Build GRCh38
rs976136321 Chr.22: 29485752-29485752 on Build GRCh38
rs897212544 Chr.22: 29485884-29485884 on Build GRCh38
rs777766167 Chr.22: 29485741-29485741 on Build GRCh38
rs1033200120 Chr.22: 29485821-29485821 on Build GRCh38
rs148653339 Chr.22: 29485808-29485808 on Build GRCh38
rs267607674 Chr.22: 29485842-29485842 on Build GRCh38
rs750653120 Chr.22: 29485871-29485876 on Build GRCh38
rs796263864 Chr.22: 29485892-29485892 on Build GRCh38
rs777819212 Chr.22: 29485829-29485829 on Build GRCh38
rs533560885 Chr.22: 29485823-29485823 on Build GRCh38
rs143500523 Chr.22: 29485888-29485888 on Build GRCh38
rs746775470 Chr.22: 29485742-29485742 on Build GRCh38
rs768916813 Chr.22: 29485849-29485849 on Build GRCh38
rs372676194 Chr.22: 29485776-29485776 on Build GRCh38
rs765917980 Chr.22: 29485690-29485690 on Build GRCh38
rs199907249 Chr.22: 29485744-29485744 on Build GRCh38
rs758408756 Chr.22: 29485715-29485715 on Build GRCh38
rs764849833 Chr.22: 29485780-29485780 on Build GRCh38
rs766716879 Chr.22: 29485789-29485789 on Build GRCh38
rs750780165 Chr.22: 29485677-29485677 on Build GRCh38
rs935124849 Chr.22: 29485745-29485745 on Build GRCh38
rs201416955 Chr.22: 29485777-29485777 on Build GRCh38
rs369411473 Chr.22: 29485854-29485854 on Build GRCh38
rs187983020 Chr.22: 29485874-29485874 on Build GRCh38
rs758456135 Chr.22: 29485828-29485828 on Build GRCh38
rs753489717 Chr.22: 29485693-29485693 on Build GRCh38
rs759136568 Chr.22: 29485726-29485726 on Build GRCh38
rs540168753 Chr.22: 29485896-29485896 on Build GRCh38
rs1036179826 Chr.22: 29485883-29485883 on Build GRCh38
rs764910272 Chr.22: 29485819-29485819 on Build GRCh38
rs4823040 Chr.22: 29485895-29485895 on Build GRCh38
rs117036372 Chr.22: 29485855-29485855 on Build GRCh38
rs768630912 Chr.22: 29485775-29485775 on Build GRCh38
rs867469071 Chr.22: 29485755-29485755 on Build GRCh38
rs764959146 Chr.22: 29485887-29485887 on Build GRCh38
rs748104186 Chr.22: 29485707-29485707 on Build GRCh38
rs754469100 Chr.22: 29485697-29485697 on Build GRCh38
rs144321945 Chr.22: 29485832-29485832 on Build GRCh38
rs770091029 Chr.22: 29485743-29485743 on Build GRCh38
rs749476008 Chr.22: 29485845-29485845 on Build GRCh38
rs773730940 Chr.22: 29485861-29485861 on Build GRCh38
rs759172742 Chr.22: 29485885-29485885 on Build GRCh38
rs776382146 Chr.22: 29485678-29485678 on Build GRCh38
rs755841191 Chr.22: 29485684-29485684 on Build GRCh38
rs377149236 Chr.22: 29485787-29485787 on Build GRCh38
rs968210046 Chr.22: 29485732-29485732 on Build GRCh38
rs774469755 Chr.22: 29485851-29485851 on Build GRCh38
rs753450146 Chr.22: 29485801-29485801 on Build GRCh38
rs761187527 Chr.22: 29485872-29485872 on Build GRCh38
rs373516936 Chr.22: 29485698-29485698 on Build GRCh38
rs529431783 Chr.22: 29485897-29485897 on Build GRCh38

View Full Product Details