Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100635053-100635324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN6
Entrez Gene ID
7105
Gene Name
tetraspanin 6
Gene Aliases
T245, TM4SF6, TSPAN-6
UniGene
Hs.43233
Gene Chromosome Location
Chr.X: 100627108-100637104 on Build GRCh38
Transcripts
NM_001278740.1 NM_001278741.1 NM_001278742.1 NM_001278743.1 NM_003270.3 XM_011531018.1

SNP Detail

SNP ID Chromosome Location
rs765693066 Chr.X: 100635252-100635252 on Build GRCh38
rs757907711 Chr.X: 100635224-100635224 on Build GRCh38
rs748564677 Chr.X: 100635178-100635178 on Build GRCh38
rs111978433 Chr.X: 100635103-100635103 on Build GRCh38
rs141301339 Chr.X: 100635229-100635229 on Build GRCh38
rs1802288 Chr.X: 100635207-100635207 on Build GRCh38
rs200238785 Chr.X: 100635210-100635210 on Build GRCh38
rs753564797 Chr.X: 100635205-100635205 on Build GRCh38
rs369119853 Chr.X: 100635219-100635219 on Build GRCh38
rs756394862 Chr.X: 100635246-100635246 on Build GRCh38
rs146592098 Chr.X: 100635211-100635211 on Build GRCh38
rs752047454 Chr.X: 100635220-100635220 on Build GRCh38
rs974606991 Chr.X: 100635202-100635202 on Build GRCh38
rs769245632 Chr.X: 100635146-100635146 on Build GRCh38
rs753996636 Chr.X: 100635264-100635264 on Build GRCh38
rs979714872 Chr.X: 100635262-100635265 on Build GRCh38
rs754962694 Chr.X: 100635268-100635268 on Build GRCh38
rs199738420 Chr.X: 100635296-100635296 on Build GRCh38
rs766479071 Chr.X: 100635256-100635256 on Build GRCh38
rs762341104 Chr.X: 100635220-100635220 on Build GRCh38
rs373921492 Chr.X: 100635244-100635244 on Build GRCh38
rs764529766 Chr.X: 100635201-100635201 on Build GRCh38
rs761628915 Chr.X: 100635241-100635241 on Build GRCh38
rs2234089 Chr.X: 100635261-100635261 on Build GRCh38
rs779502112 Chr.X: 100635279-100635279 on Build GRCh38
rs377508757 Chr.X: 100635145-100635145 on Build GRCh38
rs779264805 Chr.X: 100635156-100635156 on Build GRCh38
rs750589911 Chr.X: 100635294-100635294 on Build GRCh38
rs775136588 Chr.X: 100635213-100635213 on Build GRCh38

View Full Product Details