Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62981063-62981316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A6
Entrez Gene ID
9356
Gene Name
solute carrier family 22 member 6
Gene Aliases
HOAT1, OAT1, PAHT, ROAT1
UniGene
Hs.369252
Gene Chromosome Location
Chr.11: 62976597-62985023 on Build GRCh38
Transcripts
NM_004790.4 NM_153276.2 NM_153277.2 NM_153278.2 XM_017018562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4973624 c.888G>A p.G296G Chr.11: 62981293-62981293 on Build GRCh38
7208812 c.955A>T p.M319L Chr.11: 62981067-62981067 on Build GRCh38
6780303 c.883A>G p.N295D Chr.11: 62981298-62981298 on Build GRCh38
4726727 c.873C>T p.V291V Chr.11: 62981308-62981308 on Build GRCh38
4561769 c.898G>A p.E300K Chr.11: 62981283-62981283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762648329 Chr.11: 62981093-62981093 on Build GRCh38
rs1027075922 Chr.11: 62981114-62981114 on Build GRCh38
rs901177430 Chr.11: 62981211-62981211 on Build GRCh38
rs766433884 Chr.11: 62981284-62981284 on Build GRCh38
rs535236835 Chr.11: 62981212-62981212 on Build GRCh38
rs2276300 Chr.11: 62981227-62981227 on Build GRCh38
rs774014383 Chr.11: 62981247-62981247 on Build GRCh38
rs776644088 Chr.11: 62981288-62981288 on Build GRCh38
rs868659836 Chr.11: 62981177-62981177 on Build GRCh38
rs756766644 Chr.11: 62981280-62981282 on Build GRCh38
rs761626832 Chr.11: 62981111-62981111 on Build GRCh38
rs766549276 Chr.11: 62981122-62981122 on Build GRCh38
rs931445013 Chr.11: 62981118-62981118 on Build GRCh38
rs539267696 Chr.11: 62981116-62981116 on Build GRCh38
rs199801650 Chr.11: 62981282-62981282 on Build GRCh38
rs370684842 Chr.11: 62981275-62981275 on Build GRCh38
rs11568617 Chr.11: 62981129-62981129 on Build GRCh38
rs758815321 Chr.11: 62981215-62981215 on Build GRCh38
rs530672424 Chr.11: 62981091-62981091 on Build GRCh38
rs771326938 Chr.11: 62981222-62981222 on Build GRCh38
rs566545838 Chr.11: 62981201-62981201 on Build GRCh38
rs940115177 Chr.11: 62981187-62981187 on Build GRCh38
rs149591782 Chr.11: 62981271-62981271 on Build GRCh38
rs200212061 Chr.11: 62981112-62981112 on Build GRCh38
rs747615968 Chr.11: 62981220-62981220 on Build GRCh38
rs146077034 Chr.11: 62981289-62981289 on Build GRCh38
rs970580718 Chr.11: 62981206-62981206 on Build GRCh38
rs761347738 Chr.11: 62981264-62981264 on Build GRCh38
rs746609046 Chr.11: 62981105-62981132 on Build GRCh38
rs376711110 Chr.11: 62981094-62981094 on Build GRCh38
rs748797963 Chr.11: 62981228-62981228 on Build GRCh38
rs768114265 Chr.11: 62981246-62981246 on Build GRCh38
rs914594900 Chr.11: 62981276-62981276 on Build GRCh38
rs774974401 Chr.11: 62981092-62981092 on Build GRCh38
rs369866532 Chr.11: 62981128-62981128 on Build GRCh38
rs779283504 Chr.11: 62981131-62981131 on Build GRCh38
rs1032179456 Chr.11: 62981252-62981252 on Build GRCh38

View Full Product Details