Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118373559-118373784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE4A
Entrez Gene ID
9354
Gene Name
ubiquitination factor E4A
Gene Aliases
E4, UBOX2, UFD2
UniGene
Hs.602967
Gene Chromosome Location
Chr.11: 118359581-118399211 on Build GRCh38
Transcripts
NM_001204077.1 NM_004788.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923553 c.1046C>A p.T349N Chr.11: 118373594-118373594 on Build GRCh38
7100854 c.1110C>G p.I370M Chr.11: 118373658-118373658 on Build GRCh38
28475 c.1025G>T p.S342I Chr.11: 118373573-118373573 on Build GRCh38
923552 c.1019T>G p.I340S Chr.11: 118373567-118373567 on Build GRCh38
4987544 c.1028T>C p.I343T Chr.11: 118373576-118373576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773081224 Chr.11: 118373659-118373659 on Build GRCh38
rs907156805 Chr.11: 118373638-118373638 on Build GRCh38
rs757387623 Chr.11: 118373622-118373622 on Build GRCh38
rs751151916 Chr.11: 118373718-118373718 on Build GRCh38
rs901151695 Chr.11: 118373765-118373765 on Build GRCh38
rs986144860 Chr.11: 118373675-118373675 on Build GRCh38
rs780223787 Chr.11: 118373632-118373632 on Build GRCh38
rs756479938 Chr.11: 118373605-118373605 on Build GRCh38
rs878974179 Chr.11: 118373724-118373724 on Build GRCh38
rs754143979 Chr.11: 118373735-118373735 on Build GRCh38
rs778910049 Chr.11: 118373627-118373627 on Build GRCh38
rs764510243 Chr.11: 118373722-118373722 on Build GRCh38
rs751268784 Chr.11: 118373596-118373596 on Build GRCh38
rs144679787 Chr.11: 118373670-118373670 on Build GRCh38
rs1006066609 Chr.11: 118373682-118373682 on Build GRCh38
rs779667541 Chr.11: 118373627-118373628 on Build GRCh38
rs961512592 Chr.11: 118373750-118373750 on Build GRCh38
rs769625932 Chr.11: 118373658-118373658 on Build GRCh38
rs368222607 Chr.11: 118373674-118373674 on Build GRCh38
rs1010099057 Chr.11: 118373618-118373618 on Build GRCh38
rs377727078 Chr.11: 118373681-118373681 on Build GRCh38
rs1021612573 Chr.11: 118373709-118373709 on Build GRCh38
rs754370282 Chr.11: 118373617-118373617 on Build GRCh38
rs752955616 Chr.11: 118373602-118373602 on Build GRCh38
rs752363475 Chr.11: 118373698-118373698 on Build GRCh38
rs759286724 Chr.11: 118373676-118373676 on Build GRCh38
rs891642693 Chr.11: 118373598-118373598 on Build GRCh38
rs761014676 Chr.11: 118373719-118373719 on Build GRCh38
rs369189476 Chr.11: 118373639-118373639 on Build GRCh38
rs201361239 Chr.11: 118373587-118373587 on Build GRCh38
rs75516217 Chr.11: 118373607-118373607 on Build GRCh38
rs972910590 Chr.11: 118373751-118373751 on Build GRCh38
rs375416078 Chr.11: 118373714-118373714 on Build GRCh38
rs772230081 Chr.11: 118373629-118373629 on Build GRCh38
rs922855258 Chr.11: 118373758-118373758 on Build GRCh38
rs75571765 Chr.11: 118373628-118373628 on Build GRCh38
rs762888382 Chr.11: 118373665-118373665 on Build GRCh38
rs374969961 Chr.11: 118373597-118373597 on Build GRCh38

View Full Product Details