Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135887955-135888210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A48
Entrez Gene ID
153328
Gene Name
solute carrier family 25 member 48
Gene Aliases
-
UniGene
Hs.412418
Gene Chromosome Location
Chr.5: 135579002-135888637 on Build GRCh38
Transcripts
NM_145282.4 XM_006714550.1 XM_011543199.1 XM_017009086.1 XM_017009090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5823573 c.467G>T p.C156F Chr.5: 135888077-135888077 on Build GRCh38
6354244 c.422-8G>T p.? Chr.5: 135888024-135888024 on Build GRCh38
3428917 c.422G>A p.G141E Chr.5: 135888032-135888032 on Build GRCh38
6469941 c.452C>G p.P151R Chr.5: 135888062-135888062 on Build GRCh38
5402137 c.427G>T p.E143* Chr.5: 135888037-135888037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528581671 Chr.5: 135888139-135888139 on Build GRCh38
rs530450145 Chr.5: 135888047-135888047 on Build GRCh38
rs770752164 Chr.5: 135887993-135887993 on Build GRCh38
rs1043376296 Chr.5: 135887992-135887992 on Build GRCh38
rs1027626170 Chr.5: 135888082-135888082 on Build GRCh38
rs986141652 Chr.5: 135888065-135888065 on Build GRCh38
rs764804314 Chr.5: 135888023-135888023 on Build GRCh38
rs916771753 Chr.5: 135888173-135888173 on Build GRCh38
rs929493969 Chr.5: 135888126-135888126 on Build GRCh38
rs542503843 Chr.5: 135888072-135888072 on Build GRCh38
rs146520819 Chr.5: 135888145-135888145 on Build GRCh38
rs73791626 Chr.5: 135888085-135888085 on Build GRCh38
rs995847426 Chr.5: 135888081-135888081 on Build GRCh38
rs9327734 Chr.5: 135888024-135888024 on Build GRCh38
rs1020823348 Chr.5: 135888008-135888008 on Build GRCh38
rs547302414 Chr.5: 135888144-135888144 on Build GRCh38
rs780102872 Chr.5: 135888103-135888103 on Build GRCh38
rs202001446 Chr.5: 135888073-135888073 on Build GRCh38
rs746766274 Chr.5: 135887982-135887982 on Build GRCh38
rs113880946 Chr.5: 135888045-135888045 on Build GRCh38
rs73287302 Chr.5: 135888080-135888080 on Build GRCh38
rs768100399 Chr.5: 135888032-135888032 on Build GRCh38
rs762394386 Chr.5: 135888030-135888030 on Build GRCh38
rs112481078 Chr.5: 135888109-135888109 on Build GRCh38
rs766904264 Chr.5: 135888050-135888050 on Build GRCh38
rs759299701 Chr.5: 135888013-135888013 on Build GRCh38
rs991998720 Chr.5: 135888006-135888006 on Build GRCh38
rs990969873 Chr.5: 135888156-135888156 on Build GRCh38

View Full Product Details