Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67748569-67748814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE26
Entrez Gene ID
23503
Gene Name
zinc finger FYVE-type containing 26
Gene Aliases
FYVE-CENT, SPG15
UniGene
Hs.98041
Gene Chromosome Location
Chr.14: 67728892-67817293 on Build GRCh38
Transcripts
NM_015346.3 XM_017021124.1 XM_017021125.1 XM_017021126.1 XM_017021127.1 XM_017021128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7148153 c.7477G>C p.E2493Q Chr.14: 67748579-67748579 on Build GRCh38
6001261 c.7417-8A>G p.? Chr.14: 67748647-67748647 on Build GRCh38
5694102 c.7466C>T p.A2489V Chr.14: 67748590-67748590 on Build GRCh38
957217 c.7460T>C p.L2487S Chr.14: 67748596-67748596 on Build GRCh38
4942718 c.7450T>C p.S2484P Chr.14: 67748606-67748606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756257064 Chr.14: 67748636-67748636 on Build GRCh38
rs372502591 Chr.14: 67748682-67748682 on Build GRCh38
rs893113637 Chr.14: 67748740-67748740 on Build GRCh38
rs1008400456 Chr.14: 67748758-67748758 on Build GRCh38
rs201854194 Chr.14: 67748609-67748609 on Build GRCh38
rs139979196 Chr.14: 67748754-67748754 on Build GRCh38
rs976321163 Chr.14: 67748593-67748593 on Build GRCh38
rs745686862 Chr.14: 67748662-67748662 on Build GRCh38
rs934705772 Chr.14: 67748718-67748718 on Build GRCh38
rs1057048372 Chr.14: 67748778-67748778 on Build GRCh38
rs181180963 Chr.14: 67748624-67748624 on Build GRCh38
rs752814396 Chr.14: 67748635-67748635 on Build GRCh38
rs939596120 Chr.14: 67748756-67748756 on Build GRCh38
rs780878664 Chr.14: 67748642-67748642 on Build GRCh38
rs372540351 Chr.14: 67748647-67748647 on Build GRCh38
rs755270814 Chr.14: 67748621-67748621 on Build GRCh38
rs201771769 Chr.14: 67748644-67748644 on Build GRCh38
rs1045374775 Chr.14: 67748750-67748750 on Build GRCh38
rs757154239 Chr.14: 67748651-67748651 on Build GRCh38
rs185269926 Chr.14: 67748775-67748775 on Build GRCh38
rs914263501 Chr.14: 67748677-67748677 on Build GRCh38
rs1053060472 Chr.14: 67748720-67748720 on Build GRCh38
rs763370008 Chr.14: 67748597-67748597 on Build GRCh38
rs948365135 Chr.14: 67748749-67748749 on Build GRCh38
rs200353381 Chr.14: 67748665-67748665 on Build GRCh38
rs766789454 Chr.14: 67748598-67748598 on Build GRCh38
rs201729037 Chr.14: 67748664-67748664 on Build GRCh38
rs778924055 Chr.14: 67748659-67748659 on Build GRCh38
rs747768786 Chr.14: 67748644-67748660 on Build GRCh38
rs755733053 Chr.14: 67748655-67748656 on Build GRCh38
rs984729057 Chr.14: 67748628-67748628 on Build GRCh38

View Full Product Details