Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189572777-189573036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC40A1
Entrez Gene ID
30061
Gene Name
solute carrier family 40 member 1
Gene Aliases
FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3
UniGene
Hs.643005
Gene Chromosome Location
Chr.2: 189560590-189580811 on Build GRCh38
Transcripts
NM_014585.5 XM_005246505.1 XM_017003938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1404040 c.381G>A p.W127* Chr.2: 189572852-189572852 on Build GRCh38
6235534 c.319G>T p.G107* Chr.2: 189572914-189572914 on Build GRCh38
3575504 c.376G>A p.G126R Chr.2: 189572857-189572857 on Build GRCh38
3990923 c.382G>T p.V128F Chr.2: 189572851-189572851 on Build GRCh38
3575503 c.377G>A p.G126E Chr.2: 189572856-189572856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201485374 Chr.2: 189572927-189572927 on Build GRCh38
rs143371720 Chr.2: 189572939-189572939 on Build GRCh38
rs911996313 Chr.2: 189572832-189572832 on Build GRCh38
rs766466332 Chr.2: 189572834-189572834 on Build GRCh38
rs1052452054 Chr.2: 189572858-189572858 on Build GRCh38
rs527680959 Chr.2: 189572826-189572826 on Build GRCh38
rs755671419 Chr.2: 189572880-189572880 on Build GRCh38
rs752118206 Chr.2: 189573004-189573004 on Build GRCh38
rs540742265 Chr.2: 189572922-189572922 on Build GRCh38
rs754108561 Chr.2: 189572835-189572835 on Build GRCh38
rs745342705 Chr.2: 189572875-189572875 on Build GRCh38
rs766487715 Chr.2: 189572967-189572967 on Build GRCh38
rs370057411 Chr.2: 189572866-189572866 on Build GRCh38
rs886055361 Chr.2: 189572944-189572944 on Build GRCh38
rs764537695 Chr.2: 189572990-189572990 on Build GRCh38
rs765493686 Chr.2: 189572986-189572986 on Build GRCh38
rs11568345 Chr.2: 189572906-189572906 on Build GRCh38
rs568086191 Chr.2: 189572860-189572860 on Build GRCh38
rs544044469 Chr.2: 189572989-189572989 on Build GRCh38
rs962732807 Chr.2: 189573002-189573002 on Build GRCh38
rs535193686 Chr.2: 189572885-189572885 on Build GRCh38
rs753119904 Chr.2: 189572859-189572859 on Build GRCh38
rs905096892 Chr.2: 189572904-189572904 on Build GRCh38
rs765633684 Chr.2: 189572851-189572851 on Build GRCh38
rs776645173 Chr.2: 189572970-189572970 on Build GRCh38
rs11568344 Chr.2: 189572846-189572846 on Build GRCh38
rs187227844 Chr.2: 189572987-189572987 on Build GRCh38
rs781723706 Chr.2: 189572986-189572990 on Build GRCh38
rs773310265 Chr.2: 189572948-189572948 on Build GRCh38
rs771203234 Chr.2: 189572975-189572975 on Build GRCh38
rs772100835 Chr.2: 189572942-189572942 on Build GRCh38

View Full Product Details