Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 136043963-136044224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFBI
Entrez Gene ID
7045
Gene Name
transforming growth factor beta induced
Gene Aliases
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
UniGene
Hs.369397
Gene Chromosome Location
Chr.5: 136028895-136063818 on Build GRCh38
Transcripts
NM_000358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454111 c.246C>T p.Y82Y Chr.5: 136044070-136044070 on Build GRCh38
1061034 c.234-2_234-1delAG p.? Chr.5: 136044056-136044057 on Build GRCh38
6436778 c.251G>T p.C84F Chr.5: 136044075-136044075 on Build GRCh38
481957 c.247G>A p.E83K Chr.5: 136044071-136044071 on Build GRCh38
6419434 c.296C>T p.A99V Chr.5: 136044120-136044120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907121717 Chr.5: 136044144-136044144 on Build GRCh38
rs370117299 Chr.5: 136044064-136044064 on Build GRCh38
rs770218208 Chr.5: 136044112-136044112 on Build GRCh38
rs929892145 Chr.5: 136044131-136044131 on Build GRCh38
rs774297092 Chr.5: 136044031-136044031 on Build GRCh38
rs958744360 Chr.5: 136043987-136043987 on Build GRCh38
rs531607032 Chr.5: 136044149-136044149 on Build GRCh38
rs748881539 Chr.5: 136044055-136044055 on Build GRCh38
rs753661856 Chr.5: 136044162-136044162 on Build GRCh38
rs574026154 Chr.5: 136044138-136044138 on Build GRCh38
rs755967823 Chr.5: 136044038-136044038 on Build GRCh38
rs779775063 Chr.5: 136044160-136044160 on Build GRCh38
rs1045682990 Chr.5: 136044132-136044132 on Build GRCh38
rs985530214 Chr.5: 136043993-136043993 on Build GRCh38
rs777335010 Chr.5: 136044079-136044079 on Build GRCh38
rs759371406 Chr.5: 136044184-136044184 on Build GRCh38
rs1022470598 Chr.5: 136044185-136044185 on Build GRCh38
rs779826356 Chr.5: 136044045-136044045 on Build GRCh38
rs764412216 Chr.5: 136044129-136044129 on Build GRCh38
rs373032754 Chr.5: 136044166-136044166 on Build GRCh38
rs145627157 Chr.5: 136044005-136044005 on Build GRCh38
rs768375107 Chr.5: 136044058-136044058 on Build GRCh38
rs373533246 Chr.5: 136044070-136044070 on Build GRCh38
rs1034184060 Chr.5: 136043983-136043983 on Build GRCh38
rs776013046 Chr.5: 136044117-136044117 on Build GRCh38
rs939344125 Chr.5: 136044155-136044155 on Build GRCh38
rs902715663 Chr.5: 136044191-136044191 on Build GRCh38
rs549818448 Chr.5: 136044174-136044174 on Build GRCh38
rs886059922 Chr.5: 136044107-136044107 on Build GRCh38
rs771320956 Chr.5: 136044074-136044074 on Build GRCh38
rs376035494 Chr.5: 136044171-136044171 on Build GRCh38
rs770567633 Chr.5: 136044028-136044028 on Build GRCh38
rs759976271 Chr.5: 136044097-136044097 on Build GRCh38
rs370812166 Chr.5: 136044024-136044024 on Build GRCh38
rs370840238 Chr.5: 136044137-136044137 on Build GRCh38
rs548789955 Chr.5: 136044080-136044080 on Build GRCh38
rs377463552 Chr.5: 136044123-136044123 on Build GRCh38

View Full Product Details