Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222727069-222727342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BROX
Entrez Gene ID
148362
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
C1orf58
UniGene
Hs.534965
Gene Chromosome Location
Chr.1: 222712553-222735196 on Build GRCh38
Transcripts
NM_001288579.1 NM_001288580.1 NM_001288581.1 NM_144695.3 XM_005273065.2 XM_005273069.4 XM_006711173.3 XM_011509212.2 XM_011509213.2 XM_011509214.2 XM_017000374.1 XM_017000375.1 XM_017000376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1339097 c.593G>A p.R198Q Chr.1: 222727180-222727180 on Build GRCh38
6061745 c.656T>C p.F219S Chr.1: 222727243-222727243 on Build GRCh38
6612898 c.662delA p.A223fs*22 Chr.1: 222727249-222727249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1043273318 Chr.1: 222727119-222727119 on Build GRCh38
rs767165764 Chr.1: 222727250-222727250 on Build GRCh38
rs369931734 Chr.1: 222727276-222727276 on Build GRCh38
rs763833733 Chr.1: 222727290-222727290 on Build GRCh38
rs369688284 Chr.1: 222727289-222727289 on Build GRCh38
rs544801453 Chr.1: 222727281-222727281 on Build GRCh38
rs755548693 Chr.1: 222727258-222727258 on Build GRCh38
rs752490391 Chr.1: 222727237-222727237 on Build GRCh38
rs766330609 Chr.1: 222727132-222727132 on Build GRCh38
rs5023666 Chr.1: 222727222-222727222 on Build GRCh38
rs756758948 Chr.1: 222727247-222727247 on Build GRCh38
rs774941919 Chr.1: 222727292-222727293 on Build GRCh38
rs533643203 Chr.1: 222727281-222727281 on Build GRCh38
rs760790132 Chr.1: 222727203-222727203 on Build GRCh38
rs201017265 Chr.1: 222727207-222727207 on Build GRCh38
rs769095263 Chr.1: 222727188-222727188 on Build GRCh38
rs374393564 Chr.1: 222727277-222727277 on Build GRCh38
rs749849840 Chr.1: 222727185-222727185 on Build GRCh38
rs765045661 Chr.1: 222727156-222727156 on Build GRCh38
rs754340925 Chr.1: 222727253-222727253 on Build GRCh38
rs371178531 Chr.1: 222727246-222727246 on Build GRCh38
rs745734666 Chr.1: 222727282-222727282 on Build GRCh38
rs762718799 Chr.1: 222727240-222727240 on Build GRCh38
rs753825180 Chr.1: 222727135-222727135 on Build GRCh38
rs779632079 Chr.1: 222727268-222727268 on Build GRCh38
rs763788766 Chr.1: 222727243-222727243 on Build GRCh38
rs759298448 Chr.1: 222727137-222727137 on Build GRCh38
rs201590734 Chr.1: 222727097-222727104 on Build GRCh38
rs147619457 Chr.1: 222727173-222727173 on Build GRCh38
rs780586781 Chr.1: 222727180-222727180 on Build GRCh38
rs536323564 Chr.1: 222727193-222727193 on Build GRCh38
rs779434173 Chr.1: 222727189-222727189 on Build GRCh38
rs773368101 Chr.1: 222727198-222727198 on Build GRCh38
rs529267261 Chr.1: 222727179-222727179 on Build GRCh38
rs1027592761 Chr.1: 222727309-222727309 on Build GRCh38
rs770959156 Chr.1: 222727205-222727205 on Build GRCh38
rs774443498 Chr.1: 222727183-222727183 on Build GRCh38
rs1029267914 Chr.1: 222727136-222727136 on Build GRCh38
rs147747663 Chr.1: 222727304-222727304 on Build GRCh38
rs748510757 Chr.1: 222727270-222727270 on Build GRCh38
rs372955065 Chr.1: 222727225-222727225 on Build GRCh38
rs377395947 Chr.1: 222727279-222727279 on Build GRCh38
rs748314648 Chr.1: 222727192-222727192 on Build GRCh38
rs764950848 Chr.1: 222727226-222727226 on Build GRCh38
rs758080144 Chr.1: 222727163-222727163 on Build GRCh38
rs796840595 Chr.1: 222727102-222727102 on Build GRCh38
rs759411087 Chr.1: 222727279-222727280 on Build GRCh38
rs769710279 Chr.1: 222727285-222727285 on Build GRCh38
rs752294333 Chr.1: 222727161-222727161 on Build GRCh38

View Full Product Details