Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 686640-686883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR24
Entrez Gene ID
84219
Gene Name
WD repeat domain 24
Gene Aliases
C16orf21, JFP7
UniGene
Hs.459632
Gene Chromosome Location
Chr.16: 684622-690661 on Build GRCh38
Transcripts
NM_032259.3 XM_011522699.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731901 c.1632C>G p.D544E Chr.16: 686834-686834 on Build GRCh38
3717071 c.1617G>T p.M539I Chr.16: 686849-686849 on Build GRCh38
7337758 c.1219G>A p.G407S Chr.16: 686857-686857 on Build GRCh38
5637376 c.1591G>A p.E531K Chr.16: 686875-686875 on Build GRCh38
7348683 c.1312C>T p.R438* Chr.16: 686764-686764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751802981 Chr.16: 686771-686771 on Build GRCh38
rs770697939 Chr.16: 686673-686673 on Build GRCh38
rs747883266 Chr.16: 686815-686815 on Build GRCh38
rs947694016 Chr.16: 686671-686671 on Build GRCh38
rs1010397831 Chr.16: 686776-686776 on Build GRCh38
rs765066319 Chr.16: 686823-686823 on Build GRCh38
rs921203779 Chr.16: 686837-686837 on Build GRCh38
rs756705061 Chr.16: 686795-686795 on Build GRCh38
rs375705950 Chr.16: 686858-686858 on Build GRCh38
rs777303370 Chr.16: 686859-686859 on Build GRCh38
rs906195666 Chr.16: 686692-686692 on Build GRCh38
rs553718342 Chr.16: 686680-686680 on Build GRCh38
rs370077644 Chr.16: 686862-686862 on Build GRCh38
rs756149411 Chr.16: 686850-686850 on Build GRCh38
rs572132458 Chr.16: 686681-686681 on Build GRCh38
rs926079834 Chr.16: 686785-686786 on Build GRCh38
rs753690080 Chr.16: 686751-686751 on Build GRCh38
rs752407669 Chr.16: 686824-686824 on Build GRCh38
rs760706058 Chr.16: 686731-686731 on Build GRCh38
rs770451789 Chr.16: 686860-686860 on Build GRCh38
rs759421061 Chr.16: 686819-686819 on Build GRCh38
rs779458750 Chr.16: 686855-686855 on Build GRCh38
rs764978403 Chr.16: 686766-686766 on Build GRCh38
rs761327376 Chr.16: 686722-686722 on Build GRCh38
rs891944164 Chr.16: 686763-686763 on Build GRCh38
rs201947983 Chr.16: 686708-686708 on Build GRCh38
rs772843694 Chr.16: 686816-686816 on Build GRCh38
rs772574567 Chr.16: 686730-686730 on Build GRCh38
rs73487498 Chr.16: 686678-686678 on Build GRCh38
rs756250283 Chr.16: 686734-686737 on Build GRCh38
rs140632856 Chr.16: 686810-686810 on Build GRCh38
rs777855562 Chr.16: 686780-686782 on Build GRCh38
rs987559212 Chr.16: 686839-686839 on Build GRCh38
rs146799765 Chr.16: 686794-686794 on Build GRCh38
rs201523704 Chr.16: 686724-686724 on Build GRCh38
rs200295900 Chr.16: 686854-686854 on Build GRCh38
rs749743422 Chr.16: 686805-686805 on Build GRCh38
rs150401929 Chr.16: 686848-686848 on Build GRCh38
rs766220895 Chr.16: 686739-686739 on Build GRCh38
rs377320675 Chr.16: 686847-686847 on Build GRCh38
rs957789410 Chr.16: 686789-686789 on Build GRCh38
rs374231595 Chr.16: 686817-686817 on Build GRCh38
rs780238415 Chr.16: 686852-686852 on Build GRCh38
rs372812017 Chr.16: 686857-686857 on Build GRCh38
rs754414399 Chr.16: 686853-686853 on Build GRCh38
rs1023673885 Chr.16: 686713-686713 on Build GRCh38
rs367984304 Chr.16: 686707-686707 on Build GRCh38
rs754773220 Chr.16: 686753-686753 on Build GRCh38
rs780722759 Chr.16: 686801-686801 on Build GRCh38
rs932556529 Chr.16: 686846-686846 on Build GRCh38
rs77763943 Chr.16: 686777-686777 on Build GRCh38
rs151322294 Chr.16: 686806-686806 on Build GRCh38
rs757398841 Chr.16: 686774-686774 on Build GRCh38

View Full Product Details