Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153982219-153982466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM187
Entrez Gene ID
8269
Gene Name
transmembrane protein 187
Gene Aliases
CXorf12, DXS9878E, ITBA1
UniGene
Hs.23119
Gene Chromosome Location
Chr.X: 153972540-153983195 on Build GRCh38
Transcripts
NM_003492.2 XM_011531199.2

SNP Detail

SNP ID Chromosome Location
rs782401604 Chr.X: 153982257-153982257 on Build GRCh38
rs1045898 Chr.X: 153982433-153982433 on Build GRCh38
rs782741153 Chr.X: 153982273-153982273 on Build GRCh38
rs782077538 Chr.X: 153982330-153982330 on Build GRCh38
rs782264046 Chr.X: 153982386-153982386 on Build GRCh38
rs782516214 Chr.X: 153982283-153982283 on Build GRCh38
rs782508666 Chr.X: 153982387-153982387 on Build GRCh38
rs782106515 Chr.X: 153982288-153982288 on Build GRCh38
rs1039770732 Chr.X: 153982382-153982382 on Build GRCh38
rs782790890 Chr.X: 153982265-153982265 on Build GRCh38
rs2266890 Chr.X: 153982271-153982271 on Build GRCh38
rs782426347 Chr.X: 153982299-153982299 on Build GRCh38
rs376312251 Chr.X: 153982287-153982287 on Build GRCh38
rs7350355 Chr.X: 153982294-153982294 on Build GRCh38
rs147712258 Chr.X: 153982420-153982420 on Build GRCh38
rs998941791 Chr.X: 153982253-153982253 on Build GRCh38
rs782614697 Chr.X: 153982282-153982282 on Build GRCh38
rs144243296 Chr.X: 153982256-153982256 on Build GRCh38
rs782084731 Chr.X: 153982240-153982240 on Build GRCh38
rs781817194 Chr.X: 153982282-153982282 on Build GRCh38
rs782773674 Chr.X: 153982348-153982348 on Build GRCh38
rs782576612 Chr.X: 153982291-153982291 on Build GRCh38
rs927833147 Chr.X: 153982303-153982303 on Build GRCh38
rs201763538 Chr.X: 153982292-153982292 on Build GRCh38
rs782432778 Chr.X: 153982369-153982369 on Build GRCh38
rs782134915 Chr.X: 153982318-153982318 on Build GRCh38
rs781864574 Chr.X: 153982395-153982395 on Build GRCh38
rs782287239 Chr.X: 153982390-153982390 on Build GRCh38
rs782131352 Chr.X: 153982441-153982441 on Build GRCh38
rs113580872 Chr.X: 153982374-153982374 on Build GRCh38
rs782547125 Chr.X: 153982356-153982356 on Build GRCh38
rs782796110 Chr.X: 153982445-153982445 on Build GRCh38
rs200493035 Chr.X: 153982298-153982298 on Build GRCh38
rs373529673 Chr.X: 153982391-153982391 on Build GRCh38
rs149950630 Chr.X: 153982246-153982246 on Build GRCh38
rs377534936 Chr.X: 153982396-153982396 on Build GRCh38
rs782720109 Chr.X: 153982310-153982310 on Build GRCh38
rs782784538 Chr.X: 153982357-153982357 on Build GRCh38
rs782494978 Chr.X: 153982446-153982446 on Build GRCh38
rs782611312 Chr.X: 153982380-153982380 on Build GRCh38
rs782637202 Chr.X: 153982284-153982284 on Build GRCh38
rs782064943 Chr.X: 153982431-153982431 on Build GRCh38
rs370996635 Chr.X: 153982407-153982407 on Build GRCh38
rs782414882 Chr.X: 153982416-153982416 on Build GRCh38
rs142270595 Chr.X: 153982293-153982293 on Build GRCh38
rs781974966 Chr.X: 153982324-153982324 on Build GRCh38
rs782069564 Chr.X: 153982309-153982309 on Build GRCh38
rs781834823 Chr.X: 153982440-153982440 on Build GRCh38
rs782780301 Chr.X: 153982349-153982349 on Build GRCh38
rs782706680 Chr.X: 153982434-153982434 on Build GRCh38
rs782444582 Chr.X: 153982272-153982272 on Build GRCh38

View Full Product Details