Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148315586-148315813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102546294
Entrez Gene ID
102546294
Gene Name
uncharacterized LOC102546294
Gene Aliases
-
UniGene
Hs.660461
Gene Chromosome Location
Chr.5: 148268307-148383850 on Build GRCh38
Transcripts
NR_105057.1 NM_032566.2

Gene Symbol
SPINK7
Entrez Gene ID
84651
Gene Name
serine peptidase inhibitor, Kazal type 7 (putative)
Gene Aliases
ECG2, ECRG2
UniGene
Hs.244569
Gene Chromosome Location
Chr.5: 148312427-148315919 on Build GRCh38
Transcripts
NR_105057.1 NM_032566.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3211011 c.223G>A p.G75R Chr.5: 148315649-148315649 on Build GRCh38
7191075 c.221A>G p.N74S Chr.5: 148315647-148315647 on Build GRCh38
5577769 c.224G>A p.G75E Chr.5: 148315650-148315650 on Build GRCh38
1064022 c.215A>T p.K72I Chr.5: 148315641-148315641 on Build GRCh38
7391781 c.229G>A p.V77I Chr.5: 148315655-148315655 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755910109 Chr.5: 148315670-148315670 on Build GRCh38
rs773449097 Chr.5: 148315727-148315727 on Build GRCh38
rs3217238 Chr.5: 148315720-148315721 on Build GRCh38
rs541542528 Chr.5: 148315673-148315673 on Build GRCh38
rs778313840 Chr.5: 148315688-148315688 on Build GRCh38
rs147459281 Chr.5: 148315717-148315717 on Build GRCh38
rs901619725 Chr.5: 148315687-148315687 on Build GRCh38
rs753780704 Chr.5: 148315679-148315679 on Build GRCh38
rs763300167 Chr.5: 148315653-148315653 on Build GRCh38
rs564051363 Chr.5: 148315715-148315715 on Build GRCh38
rs202065676 Chr.5: 148315732-148315732 on Build GRCh38
rs72268739 Chr.5: 148315729-148315730 on Build GRCh38
rs779615320 Chr.5: 148315695-148315695 on Build GRCh38
rs957386841 Chr.5: 148315725-148315726 on Build GRCh38
rs757184484 Chr.5: 148315681-148315681 on Build GRCh38
rs187355255 Chr.5: 148315691-148315691 on Build GRCh38
rs543161075 Chr.5: 148315712-148315714 on Build GRCh38
rs111509018 Chr.5: 148315719-148315720 on Build GRCh38
rs545735744 Chr.5: 148315714-148315714 on Build GRCh38
rs768958114 Chr.5: 148315702-148315702 on Build GRCh38
rs374904796 Chr.5: 148315789-148315789 on Build GRCh38
rs764448945 Chr.5: 148315721-148315721 on Build GRCh38
rs747315773 Chr.5: 148315619-148315619 on Build GRCh38
rs375825161 Chr.5: 148315694-148315694 on Build GRCh38
rs568330209 Chr.5: 148315720-148315720 on Build GRCh38
rs34482862 Chr.5: 148315731-148315732 on Build GRCh38
rs769210670 Chr.5: 148315623-148315623 on Build GRCh38
rs192180759 Chr.5: 148315699-148315699 on Build GRCh38
rs372555086 Chr.5: 148315625-148315625 on Build GRCh38
rs774783885 Chr.5: 148315736-148315736 on Build GRCh38
rs995926079 Chr.5: 148315763-148315763 on Build GRCh38
rs748645219 Chr.5: 148315707-148315707 on Build GRCh38
rs752558358 Chr.5: 148315669-148315669 on Build GRCh38
rs201537711 Chr.5: 148315735-148315735 on Build GRCh38
rs147014109 Chr.5: 148315655-148315655 on Build GRCh38
rs765283593 Chr.5: 148315636-148315636 on Build GRCh38
rs147279316 Chr.5: 148315719-148315719 on Build GRCh38
rs1057021926 Chr.5: 148315634-148315634 on Build GRCh38
rs781600094 Chr.5: 148315703-148315703 on Build GRCh38
rs770295408 Chr.5: 148315709-148315709 on Build GRCh38
rs773313016 Chr.5: 148315640-148315640 on Build GRCh38
rs775837859 Chr.5: 148315713-148315714 on Build GRCh38
rs148739653 Chr.5: 148315764-148315765 on Build GRCh38

View Full Product Details