Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28085346-28085577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF165
Entrez Gene ID
7718
Gene Name
zinc finger protein 165
Gene Aliases
CT53, LD65, ZSCAN7
UniGene
Hs.55481
Gene Chromosome Location
Chr.6: 28080704-28104244 on Build GRCh38
Transcripts
NM_003447.3 XM_017011258.1 XM_017011259.1 XM_017011260.1 XM_017011262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1442784 c.76G>T p.E26* Chr.6: 28085556-28085556 on Build GRCh38
1233632 c.37T>C p.S13P Chr.6: 28085517-28085517 on Build GRCh38
741666 c.60G>A p.L20L Chr.6: 28085540-28085540 on Build GRCh38
5788273 c.49G>T p.E17* Chr.6: 28085529-28085529 on Build GRCh38
1271042 c.55C>T p.L19F Chr.6: 28085535-28085535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200660292 Chr.6: 28085488-28085488 on Build GRCh38
rs926235423 Chr.6: 28085405-28085405 on Build GRCh38
rs536595977 Chr.6: 28085413-28085413 on Build GRCh38
rs748234612 Chr.6: 28085376-28085376 on Build GRCh38
rs980293023 Chr.6: 28085375-28085375 on Build GRCh38
rs377116795 Chr.6: 28085439-28085439 on Build GRCh38
rs750857970 Chr.6: 28085495-28085495 on Build GRCh38
rs752319995 Chr.6: 28085509-28085509 on Build GRCh38
rs756445363 Chr.6: 28085500-28085500 on Build GRCh38
rs767985339 Chr.6: 28085489-28085489 on Build GRCh38
rs764112819 Chr.6: 28085467-28085467 on Build GRCh38
rs780396511 Chr.6: 28085502-28085502 on Build GRCh38
rs758017047 Chr.6: 28085551-28085551 on Build GRCh38
rs751592657 Chr.6: 28085475-28085475 on Build GRCh38
rs992908697 Chr.6: 28085542-28085542 on Build GRCh38
rs555684502 Chr.6: 28085419-28085420 on Build GRCh38

View Full Product Details