Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150988026-150988257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA9
Entrez Gene ID
8416
Gene Name
annexin A9
Gene Aliases
ANX31
UniGene
Hs.3346
Gene Chromosome Location
Chr.1: 150982023-150995638 on Build GRCh38
Transcripts
NM_003568.2 XM_011510058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5966080 c.787G>A p.G263S Chr.1: 150988180-150988180 on Build GRCh38
1236714 c.754C>T p.R252C Chr.1: 150988147-150988147 on Build GRCh38
3418027 c.779C>T p.A260V Chr.1: 150988172-150988172 on Build GRCh38
3473552 c.788G>A p.G263D Chr.1: 150988181-150988181 on Build GRCh38
4406148 c.778G>A p.A260T Chr.1: 150988171-150988171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373118397 Chr.1: 150988087-150988087 on Build GRCh38
rs752855411 Chr.1: 150988188-150988188 on Build GRCh38
rs367612287 Chr.1: 150988150-150988150 on Build GRCh38
rs767764082 Chr.1: 150988186-150988186 on Build GRCh38
rs767846739 Chr.1: 150988218-150988221 on Build GRCh38
rs746195112 Chr.1: 150988152-150988152 on Build GRCh38
rs756277622 Chr.1: 150988201-150988201 on Build GRCh38
rs752059226 Chr.1: 150988200-150988200 on Build GRCh38
rs376305826 Chr.1: 150988108-150988108 on Build GRCh38
rs587759130 Chr.1: 150988110-150988110 on Build GRCh38
rs370979889 Chr.1: 150988109-150988109 on Build GRCh38
rs766263552 Chr.1: 150988176-150988176 on Build GRCh38
rs771415277 Chr.1: 150988148-150988148 on Build GRCh38
rs776311888 Chr.1: 150988095-150988095 on Build GRCh38
rs776402629 Chr.1: 150988155-150988155 on Build GRCh38
rs766455618 Chr.1: 150988107-150988107 on Build GRCh38
rs764097530 Chr.1: 150988213-150988213 on Build GRCh38
rs749748076 Chr.1: 150988147-150988147 on Build GRCh38
rs370252595 Chr.1: 150988154-150988154 on Build GRCh38
rs761428752 Chr.1: 150988163-150988163 on Build GRCh38
rs755223923 Chr.1: 150988112-150988112 on Build GRCh38
rs373251195 Chr.1: 150988075-150988075 on Build GRCh38
rs374591781 Chr.1: 150988118-150988118 on Build GRCh38
rs77936236 Chr.1: 150988169-150988169 on Build GRCh38
rs778414328 Chr.1: 150988138-150988138 on Build GRCh38
rs149528939 Chr.1: 150988117-150988117 on Build GRCh38
rs587751896 Chr.1: 150988180-150988180 on Build GRCh38
rs147533891 Chr.1: 150988137-150988137 on Build GRCh38
rs370934834 Chr.1: 150988083-150988083 on Build GRCh38
rs778117590 Chr.1: 150988071-150988071 on Build GRCh38
rs761180071 Chr.1: 150988096-150988096 on Build GRCh38
rs1046954806 Chr.1: 150988227-150988227 on Build GRCh38
rs966589406 Chr.1: 150988146-150988146 on Build GRCh38
rs587650739 Chr.1: 150988217-150988217 on Build GRCh38
rs769172343 Chr.1: 150988165-150988165 on Build GRCh38
rs376529122 Chr.1: 150988081-150988081 on Build GRCh38
rs754385534 Chr.1: 150988125-150988125 on Build GRCh38
rs755540101 Chr.1: 150988218-150988219 on Build GRCh38
rs143896675 Chr.1: 150988101-150988101 on Build GRCh38
rs369064246 Chr.1: 150988098-150988098 on Build GRCh38
rs1028463218 Chr.1: 150988102-150988102 on Build GRCh38
rs774149401 Chr.1: 150988179-150988179 on Build GRCh38
rs762802671 Chr.1: 150988172-150988172 on Build GRCh38
rs753893020 Chr.1: 150988215-150988215 on Build GRCh38
rs869174401 Chr.1: 150988085-150988087 on Build GRCh38
rs754670778 Chr.1: 150988077-150988077 on Build GRCh38

View Full Product Details